Skip to Content
Merck
All Photos(1)

Key Documents

SML2443

Sigma-Aldrich

GLPG0974

≥98% (HPLC)

Synonym(s):

4-[[(R)-1-(Benzo[b]thiophene-3-carbonyl)-2-methyl-azetidine-2-carbonyl]-(3-chloro-benzyl)-amino]-butyric acid, GLPG 0974, GLPG-0974

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$195.00
25 MG
$773.00

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$195.00
25 MG
$773.00

About This Item

Empirical Formula (Hill Notation):
C25H25ClN2O4S
CAS Number:
Molecular Weight:
485.00
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$195.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +175 to +200°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CSC2=C1C=CC=C2)N3CC[C@]3(C)C(N(CC4=CC=CC(Cl)=C4)CCCC(O)=O)=O

InChI

1S/C25H25ClN2O4S/c1-25(11-13-28(25)23(31)20-16-33-21-9-3-2-8-19(20)21)24(32)27(12-5-10-22(29)30)15-17-6-4-7-18(26)14-17/h2-4,6-9,14,16H,5,10-13,15H2,1H3,(H,29,30)/t25-/m1/s1

InChI key

MPMKMQHJHDHPBE-RUZDIDTESA-N

Biochem/physiol Actions

GLPG0974 is an orally available, high-affinity, potent and selective free fatty acid receptor 2 (FFA2; FFAR2; GPCR43; GPR43) antagonist (hFFA2 Kd = 7.5 nM; IC50 = 9 nM; Na acetate EC80-induced Ca2+ flux in HEK293 hFFA2 transfectants) with little potency toward 55 other receptors/channels/transporters (IC50 >10 μM), including FFA3 (GPR41). GLPG0974 inhibits acetate-induced human neutrophil migration (IC50 = 27 nM) and exhibits in vivo efficacy (0.1 or 1 mg/kg intragastrically) against duodenal lesions induction by NSAID indomethacin/FFA2 agonist phenylacetamide-1 co-treatment with good pharmacokinetic properties and oral availability (F = 47% in rats; 5 mg/kg p.o.).
Orally available, high-affinity, potent and selective free fatty acid receptor 2 (FFA2; FFAR2; GPCR43; GPR43) antagonist with in vitro and in vivo efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amira Sayed Khan et al.
Cellular and molecular gastroenterology and hepatology, 15(3), 633-663 (2022-11-22)
The spontaneous preference for dietary lipids is principally regulated by 2 lingual fat taste receptors, CD36 and GPR120. Obese animals and most of human subjects exhibit low orosensory perception of dietary fat because of malfunctioning of these taste receptors. Our
Xianzhang Zeng et al.
Microbiome, 11(1), 204-204 (2023-09-12)
Aging is a significant risk factor for ischemic stroke and worsens its outcome. However, the mechanisms for this worsened neurological outcome with aging are not clearly defined. Old C57BL/6J male mice (18 to 20 months old) had a poorer neurological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service