Skip to Content
Merck
All Photos(1)

Key Documents

SML2436

Sigma-Aldrich

BAY-958 hydrochloride

≥98% (HPLC)

Synonym(s):

(3-((4-(4-Fluoro-2-methoxyphenyl)-1,3,5-triazin-2-yl)amino)phenyl)methanesulfonamide hydrochloride, 3-[[4-(4-Fluoro-2-methoxyphenyl)-1,3,5-triazin-2-yl]amino]benzenemethanesulfonamide hydrochloride, BAY 958 HCl, BAY958 HCl, LDC 526 HCl, LDC526 HCl

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$130.00
25 MG
$422.00

$130.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$130.00
25 MG
$422.00

About This Item

Empirical Formula (Hill Notation):
C17H16FN5O3S · HCl
CAS Number:
Molecular Weight:
425.86
UNSPSC Code:
12352200
NACRES:
NA.77

$130.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=CC=C(C2=NC(NC3=CC(CS(=O)(N)=O)=CC=C3)=NC=N2)C(OC)=C1.Cl

InChI key

WKPVFIKSRZVXOX-UHFFFAOYSA-N

Biochem/physiol Actions

BAY-958 (LDC526) is an orally active, ATP site-targeting, potent and selective PTEFb/CDK9 inhibitor (hCDK9-CycT1 IC50 = 5-11 nM; [ATP] = 10 μM) with 52- to >2000-fold selectivity over CDK1-7 with antiproliferative activity in cancer cultures (IC50 = 0.28 μM/96h/MOLM-13, 1 μM/96h/HeLa, 0.55 μM/48h/MEC-1, 0.28 μM/48h/CCL patient PBMCs). When administered orally in mice, BAY-958 significantly suppresses MOLM-13 acute myeloid leukemia (AML) tumor growth in mice (30/40 mg/kg/day p.o.; HCl salt) and exhibits in vivo efficacy in two murine chronic lymphocytic leukemia (CLL) models (50 & 70 mg/kg/day p.o.; free base).
Orally active, ATP site-targeting, potent and selective PTEFb/CDK9 inhibitor with anti-cancer efficacy in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ulrich Lücking et al.
ChemMedChem, 12(21), 1776-1793 (2017-09-30)
Selective inhibition of exclusively transcription-regulating PTEFb/CDK9 is a promising new approach in cancer therapy. Starting from lead compound BAY-958, lead optimization efforts strictly focusing on kinase selectivity, physicochemical and DMPK properties finally led to the identification of the orally available
Joachim R Göthert et al.
Oncotarget, 9(41), 26353-26369 (2018-06-15)
Onset of progression even during therapy with novel drugs remains an issue in chronic lymphocytic leukemia (CLL). Thus, there is ongoing demand for novel agents. Approaches targeting cyclin-dependent kinases (CDK) have reached the clinical trial stage. CDK9 mediating RNA transcriptional

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service