Skip to Content
Merck
All Photos(1)

Key Documents

SML2404

Sigma-Aldrich

EC23

≥98% (HPLC)

Synonym(s):

4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethynyl)benzoic acid, 4-[2-(5,6,7,8-Tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)ethynyl]benzoic acid, AGN 190205, AGN-190205, AGN190205, BASF 46928, BASF-46928, BASF46928, EC 23, EC-23

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$147.70
25 MG
$597.10

$147.70


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$147.70
25 MG
$597.10

About This Item

Empirical Formula (Hill Notation):
C23H24O2
CAS Number:
Molecular Weight:
332.44
MDL number:
UNSPSC Code:
12352200

$147.70


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(cc1)C#Cc2cc3c(cc2)C(CCC3(C)C)(C)C

InChI

1S/C23H24O2/c1-22(2)13-14-23(3,4)20-15-17(9-12-19(20)22)6-5-16-7-10-18(11-8-16)21(24)25/h7-12,15H,13-14H2,1-4H3,(H,24,25)

InChI key

OQVLOWLEEHYBJH-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

EC23 is a synthetic, photostable all-trans retinoic acid (ATRA) analog/mimetic that targets RAR with greater affinity than ATRA (RARα/β/γ EC50 by cell-free TR-FRET binding assay = 3.7/3.3/16.8 with EC23 vs. 16.0/17.6/14.7 nM with ATRA) and does not bind retinoid x receptors (RXRs). EC23 induces differentiation of human pluripotent embryonic stem cells (0.1-10 μM) with a similar or better efficiency than ATRA.
Photostable all-trans retinoic acid (ATRA) mimetic that targets RAR with greater affinity and induces stem cells differentiation with better efficiency than ATRA.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Maltman et al.
Molecular bioSystems, 5(5), 458-471 (2009-04-22)
The natural retinoid, all-trans retinoic acid (ATRA), is widely used to direct the in vitro differentiation of stem cells. However, substantial degradation and isomerisation of ATRA in response to UV-vis light has serious implications with regard to experimental reproducibility and
Haider M Hassan et al.
Cell reports, 19(8), 1685-1697 (2017-05-26)
Retinoic acid (RA) plays important roles in development, growth, and homeostasis through regulation of the nuclear receptors for RA (RARs). Herein, we identify Hypermethylated in Cancer 1 (Hic1) as an RA-inducible gene. HIC1 encodes a tumor suppressor, which is often
Hesham Haffez et al.
Molecular neurobiology, 55(3), 1942-1950 (2017-03-01)
All-trans retinoic acid (ATRA) plays key roles in neurogenesis mediated by retinoic acid receptors (RARs). RARs are important targets for the therapeutic regulation of neurogenesis but effective drug development depends on modelling-based strategies to design high-specificity ligands in combination with
Kirsty E Clarke et al.
Neurochemistry international, 106, 74-84 (2016-12-25)
The inability of neurites to grow and restore neural connections is common to many neurological disorders, including trauma to the central nervous system and neurodegenerative diseases. Therefore, there is need for a robust and reproducible model of neurite outgrowth, to
Victoria B Christie et al.
Organic & biomolecular chemistry, 6(19), 3497-3507 (2008-12-17)
All-trans-retinoic acid (ATRA) and its associated analogues are important mediators of cell differentiation and function during the development of the nervous system. It is well known that ATRA can induce the differentiation of neural tissues from human pluripotent stem cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service