Skip to Content
Merck
All Photos(1)

Key Documents

SML2376

Sigma-Aldrich

GSK′963

≥98% (HPLC)

Synonym(s):

1-[(5S)-4,5-Dihydro-5-phenyl-1H-pyrazol-1-yl]-2,2-dimethyl-1-propanone; 2,2-Dimethyl-1-(5(S)-phenyl-4,5-dihydro-pyrazol-1-yl)-propan-1-one, GSK 963, GSK′ 963A, GSK′963A, GSK-963, GSK-963A, GSK963, GSK963A

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$161.00
25 MG
$645.00

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$161.00
25 MG
$645.00

About This Item

Empirical Formula (Hill Notation):
C14H18N2O
CAS Number:
Molecular Weight:
230.31
UNSPSC Code:
12352200
NACRES:
NA.77

$161.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C(C)(C)C)N1[C@H](C2=CC=CC=C2)CC=N1

Biochem/physiol Actions

Brain-penetrant, highly potent and selective RIPK1 (RIP1) inhibitor with necroptosis blocking efficacy in vitro and in vivo.
GSK′963 (GSK′963A) is a brain-penetrant, highly potent and selective ATP site-targeting receptor-interacting protein 1 kinase (RIP1; RIPK1) inhibitor (IC50 = 0.8-8 nM with 50 μM ATP; IC50 >10 μM against RIPK2/3/5 and 335 other kinases) that protects against TNFα/zVAD-induced necroptosis (EC50 = 1/4 nM in mouse L929/human U937 cultures) and blocks Y. pestis-induced death of murine fetal liver macrophages (1 μM). GSK′963 prevents lethal hypothermia by acute sterile shock (2 mg/kg i.p. 15 min prior to TNFα/zVAD i.v.) and protects against acute neuronal death upon autologous blood intracerebral hemorrhage induction in mice in vivo (25 mg/kg/3 hr i.p.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sevda Lule et al.
Stroke, 48(9), 2549-2556 (2017-08-03)
Recent studies using cultured cells and rodent intracerebral hemorrhage (ICH) models have implicated RIPK1 (receptor interacting protein kinase-1) as a driver of programmed necrosis and secondary injury based on use of chemical inhibitors. However, these inhibitors have off-target effects and
Hongyan Guo et al.
Cell host & microbe, 17(2), 243-251 (2015-02-13)
Herpes simplex virus (HSV)-1 and HSV-2 are significant human pathogens causing recurrent disease. During infection, HSV modulates cell death pathways using the large subunit (R1) of ribonucleotide reductase (RR) to suppress apoptosis by binding to and blocking caspase-8. Here, we demonstrate
Anne von Mässenhausen et al.
Cell death & disease, 9(3), 359-359 (2018-03-04)
Receptor-interacting protein kinases 1 and 3 (RIPK1/3) have best been described for their role in mediating a regulated form of necrosis, referred to as necroptosis. During this process, RIPK3 phosphorylates mixed lineage kinase domain-like (MLKL) to cause plasma membrane rupture.
Mallary C Greenlee-Wacker et al.
Blood, 129(24), 3237-3244 (2017-05-06)
Community-associated methicillin-resistant Staphylococcus aureus (CA-MRSA) causes infections associated with extensive tissue damage and necrosis. In vitro, human neutrophils fed CA-MRSA lyse by an unknown mechanism that is inhibited by necrostatin-1, an allosteric inhibitor of receptor-interacting serine/threonine kinase 1 (RIPK-1). RIPK-1
Wenjing Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(7), E1475-E1484 (2018-02-01)
Tubular cell necrosis is a key histological feature of acute kidney injury (AKI). Necroptosis is a type of programed necrosis, which is executed by mixed lineage kinase domain-like protein (MLKL) upon its binding to the plasma membrane. Emerging evidence indicates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service