Skip to Content
Merck
All Photos(1)

Key Documents

SML2353

Sigma-Aldrich

GPR39-C3

≥98% (HPLC)

Synonym(s):

N-[3-Chloro-4-[[[2-(methylamino)-6-(2-pyridinyl)-4-pyrimidinyl]amino]methyl]phenyl]methanesulfonamide, TC G-1008, TC-G-1008

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$169.10
25 MG
$682.70

$169.10


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$169.10
25 MG
$682.70

About This Item

Empirical Formula (Hill Notation):
C18H19ClN6O2S
CAS Number:
Molecular Weight:
418.90
UNSPSC Code:
12352200

$169.10


Estimated to ship onApril 28, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CNC1=NC(C2=CC=CC=N2)=CC(NCC3=C(Cl)C=C(NS(C)(=O)=O)C=C3)=N1

InChI

1S/C18H19ClN6O2S/c1-20-18-23-16(15-5-3-4-8-21-15)10-17(24-18)22-11-12-6-7-13(9-14(12)19)25-28(2,26)27/h3-10,25H,11H2,1-2H3,(H2,20,22,23,24)

InChI key

DRSZMILOMUPIBJ-UHFFFAOYSA-N

Biochem/physiol Actions

GPR39-C3 (TC-G-1008) is an orally available, potent and selective potent agonist of GPR39 that increases GLP-1 secretion in mice. GPR39-C3 exhibits showing an antidepressant-like effect in the forced swim test in mice.
orally available, potent and selective potent agonist of GPR39

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service