Skip to Content
Merck
All Photos(1)

Key Documents

SML2332

Sigma-Aldrich

Dynarrestin

≥98% (HPLC)

Synonym(s):

4-(4-Diethylaminophenyl)-2-(2,4-difluoro-phenylamino)-thiazole-5-carboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$149.00
25 MG
$597.00

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$149.00
25 MG
$597.00

About This Item

Empirical Formula (Hill Notation):
C22H23F2N3O2S
CAS Number:
Molecular Weight:
431.50
UNSPSC Code:
12352200
NACRES:
NA.77

$149.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)C1=CC=C(C2=C(C(OCC)=O)SC(NC3=C(F)C=C(F)C=C3)=N2)C=C1

InChI

1S/C22H23F2N3O2S/c1-4-27(5-2)16-10-7-14(8-11-16)19-20(21(28)29-6-3)30-22(26-19)25-18-12-9-15(23)13-17(18)24/h7-13H,4-6H2,1-3H3,(H,25,26)

InChI key

UZNNDDDSGCMQOQ-UHFFFAOYSA-N

Biochem/physiol Actions

Dynarrestin is a potent and selective reversible inhibitor of cytoplasmic dyneins 1 and 2 that inhibits dynein 1-dependent microtubule binding and motility without affecting ATP hydrolysis. It inhibits endosome movement and disturbs mitosis in cells. Dynarrestin inhibits dynein 2-mediated intraflagellar transport of the cargo IFT88 and flux of Smo within cilia. Also it inhibits cancer cells proliferation downstream of Smo.
Dynarrestin is an aminothiazole derivative, which can bind to protein tyrosine phosphatase interacting protein 51 (PTPIP51). This interaction helps in regulating various signaling pathways that lead to proliferation and migration.[1] Dynarrestin blocks hedgehog (Hh)-dependent signaling in mouse and human cells.[2]
Potent and selective reversible inhibitor of cytoplasmic dyneins 1 and 2 that inhibits dynein 1-dependent microtubule binding and motility without affecting ATP hydrolysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric Dietel et al.
PloS one, 14(5), e0216642-e0216642 (2019-05-11)
LDC3/Dynarrestin, an aminothiazole derivative, is a recently developed small molecule, which binds protein tyrosine phosphatase interacting protein 51 (PTPIP51). PTPIP51 interacts with various proteins regulating different signaling pathways leading to proliferation and migration. Her2 positive breast cancer cells (SKBR3) express
Ariane Schleinitz et al.
Cell reports, 42(1), 111969-111969 (2023-01-15)
The transfer of endocytosed cargoes to lysosomes (LYSs) requires HOPS, a multiprotein complex that tethers late endosomes (LEs) to LYSs before fusion. Many proteins interact with HOPS on LEs/LYSs. However, it is not clear whether these HOPS interactors localize to
Nicholas M Zehrbach et al.
Molecular biology of the cell, 34(7), ar65-ar65 (2023-04-13)
Rab GTPase-mediated vesicle trafficking of cell surface proteins, including integrins, through endocytic and recycling pathways is important in controlling cell-extracellular matrix interactions during cell migration. The focal adhesion adaptor protein, paxillin, plays a central role in regulating adhesion dynamics and
Susanne Höing et al.
Cell chemical biology, 25(4), 357-369 (2018-02-06)
Aberrant hedgehog (Hh) signaling contributes to the pathogenesis of multiple cancers. Available inhibitors target Smoothened (Smo), which can acquire mutations causing drug resistance. Thus, compounds that inhibit Hh signaling downstream of Smo are urgently needed. We identified dynarrestin, a novel
Yunan Ye et al.
EMBO reports, 23(11), e55251-e55251 (2022-10-11)
Microtubules typically promote nuclear centring during early embryonic divisions in centrosome-containing vertebrates. In acentrosomal mouse zygotes, microtubules also centre male and female pronuclei prior to the first mitosis, this time in concert with actin. How nuclear centring is brought about

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service