Skip to Content
Merck
All Photos(1)

Key Documents

SML2325

Sigma-Aldrich

BAY-6035

≥98% (HPLC)

Synonym(s):

(2S)-1-(3-Azabicyclo[3.1.0]hexane-3-carbonyl)-N-(2-cyclopropylethyl)-2-methyl-4-oxo-2,3,4,5-tetrahydro-1H-benzo[b][1,4]diazepine-7-carboxamide, BAY 6035, BAY6035

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$137.00
25 MG
$548.00

$137.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$137.00
25 MG
$548.00

About This Item

Empirical Formula (Hill Notation):
C22H28N4O3
CAS Number:
Molecular Weight:
396.48
UNSPSC Code:
12352200
NACRES:
NA.77

$137.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C[C@H](C)N(C(N2CC(C3)C3C2)=O)C4=CC=C(C(NCCC5CC5)=O)C=C4N1

InChI

1S/C22H28N4O3/c1-13-8-20(27)24-18-10-15(21(28)23-7-6-14-2-3-14)4-5-19(18)26(13)22(29)25-11-16-9-17(16)12-25/h4-5,10,13-14,16-17H,2-3,6-9,11-12H2,1H3,(H,23,28)(H,24,27)/t13-,16?,17?/m0/s1

InChI key

CKFRXCBNKKOFGO-IGEOTXOUSA-N

Biochem/physiol Actions

BAY-6035 is a substrate-competitive, potent and selective SMYD3 inhibitor (IC50 = 88 nM by cell-free MEKK2 peptide methylation; IC50 = 70 nM against cellular MEKK2 methylation) with >100-fold selectivity over other histone methyltransferases. BAY-211 is a structural analog and the recommended inactive control (IC50 = 23 μM by cell-free MEKK2 peptide methylation) for BAY-6035. For characterization details of BAY-6035, please visit the BAY-6035 probe summary on the Structural Genomics Consortium (SGC) website.

BAY-444 is the negative control for the active probe, BAY-6035. To request a sample of the negative control from the SGC, click here.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
Substrate-competitive, potent and selective SMYD3 inhibitor with >100-fold selectivity over other histone methyltransferases.

Features and Benefits

BAY-6035 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service