Skip to Content
Merck

Skip To

SML2286

BIIL260 hydrochloride

≥98% (HPLC)

Synonym(s):

4-(3-{4-[1-(4-Hydroxy-phenyl)-1-methyl-ethyl]-phenoxymethyl}-benzyloxy)-benzamidine hydrochloride, BIIL 260 hydrochloride, BIIL-260 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$113.00
$96.05
25 mg
Check Cart for Availability
$455.00

About This Item

Empirical Formula (Hill Notation):
C30H30N2O3 · xHCl
CAS Number:
Molecular Weight:
466.57 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated

$96.05

List Price$113.00Save 15%
PRICE WITHOUT NATIONAL TAXES
Web-Only Promotion

Check Cart for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N\C(=N/[H])\c1ccc(cc1)OCc2cc(ccc2)COc3ccc(cc3)C(C)(C)c4ccc(cc4)O

InChI

1S/C30H30N2O3/c1-30(2,24-8-12-26(33)13-9-24)25-10-16-28(17-11-25)35-20-22-5-3-4-21(18-22)19-34-27-14-6-23(7-15-27)29(31)32/h3-18,33H,19-20H2,1-2H3,(H3,31,32)

InChI key

MBLJFKQACMILLC-UHFFFAOYSA-N

Biochem/physiol Actions

BIIL260 is a selective and potent leukotriene B4 (BLT1) receptor antagonist with a Ki value of 1.8 nM in human U937 cells. BIIL260 was used found to act as an inverse agonist, stabilizing the inactive state of BLT1.
Leukotriene B4 (BLT1) receptor antagonist


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters