Skip to Content
Merck
All Photos(1)

Key Documents

SML2174

Sigma-Aldrich

MS012

≥98% (HPLC)

Synonym(s):

N2-Hexyl-6,7-dimethoxy-N4-(1-methylpiperidin-4-yl)quinazoline-2,4-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00
25 MG
$493.00

$122.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$122.00
25 MG
$493.00

About This Item

Empirical Formula (Hill Notation):
C22H35N5O2
CAS Number:
Molecular Weight:
401.55
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00


Estimated to ship onApril 28, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN(CC1)CCC1NC2=C3C(C=C(OC)C(OC)=C3)=NC(NCCCCCC)=N2

InChI

1S/C22H35N5O2/c1-5-6-7-8-11-23-22-25-18-15-20(29-4)19(28-3)14-17(18)21(26-22)24-16-9-12-27(2)13-10-16/h14-16H,5-13H2,1-4H3,(H2,23,24,25,26)

InChI key

ZYNUWSFRZCRKSN-UHFFFAOYSA-N

Biochem/physiol Actions

G9a-like Protein (GLP) lysine methyl transferase Inhibitor
MS012 is a selective G9a-like Protein (GLP) lysine methyl transferase (PKMT) inhibitor with an IC50 value of 7 nM and over 140-fold selectivity for GLP over G9a and other methyl transferases tested.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Xiong et al.
Bioorganic & medicinal chemistry, 25(16), 4414-4423 (2017-07-01)
Given the high homology between the protein lysine methyltransferases G9a-like protein (GLP) and G9a, it has been challenging to develop potent and selective inhibitors for either enzyme. Recently, we reported two quinazoline compounds, MS0124 and MS012, as GLP selective inhibitors.
Yan Xiong et al.
Journal of medicinal chemistry, 60(5), 1876-1891 (2017-01-31)
G9a-like protein (GLP) and G9a are highly homologous protein lysine methyltransferases (PKMTs) sharing approximately 80% sequence identity in their catalytic domains. GLP and G9a form a heterodimer complex and catalyze mono- and dimethylation of histone H3 lysine 9 and nonhistone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service