Skip to Content
Merck
All Photos(1)

Key Documents

SML2101

Sigma-Aldrich

Fosamprenavir calcium

≥98% (HPLC)

Synonym(s):

Calcium (3S) tetrahydro-3-furanyl(1S,2R)-3-[[(4-aminophenyl)sulfonyl](isobutyl)amino]-1 -benzyl-2-(phosphonooxy)propylcarbamate, GW433908

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$251.00
25 MG
$1,000.00

$251.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$251.00
25 MG
$1,000.00

About This Item

Empirical Formula (Hill Notation):
C25H34N3O9PS · Ca
CAS Number:
Molecular Weight:
623.67
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$251.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[O-]P(O[C@H](CN(CC(C)C)S(C1=CC=C(N)C=C1)(=O)=O)[C@@H](NC(O[C@@H]2COCC2)=O)CC3=CC=CC=C3)([O-])=O.[Ca+2]

InChI

1S/C25H36N3O9PS.Ca/c1-18(2)15-28(39(33,34)22-10-8-20(26)9-11-22)16-24(37-38(30,31)32)23(14-19-6-4-3-5-7-19)27-25(29)36-21-12-13-35-17-21;/h3-11,18,21,23-24H,12-17,26H2,1-2H3,(H,27,29)(H2,30,31,32);/q;+2/p-2/t21-,23-,24+;/m0./s1

InChI key

PMDQGYMGQKTCSX-HQROKSDRSA-L

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Fosamprenavir calcium is an HIV protease inhibitor anti-retroviral. It is a water soluble phosphate ester prodrug of amprenavir, the active agent.
HIV Protease Inhibitor; Antiretroviral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa L Ross et al.
The open AIDS journal, 9, 38-44 (2015-07-15)
Treatment-emergent mutations and drug resistance were analyzed in virus from HIV-infected children meeting virologic failure (VF) criteria over 48 weeks following treatment with unboosted fosamprenavir or fosamprenavir/ritonavir-containing regimens in studies APV20002 and APV29005. Both antiretroviral therapy (ART)-naïve and ART-experienced patients
Paola Nasta et al.
HIV clinical trials, 17(3), 96-108 (2016-04-30)
Safety and tolerability evaluation of adapted dose regimens containing fosamprenavir/ritonavir (FPV/r) in HIV-infected subjects with viral hepatitis co-infection. A retrospective multicohort analysis was conducted. Subjects from three European cohorts who started FPV/r or lopinavir/ritonavir (LPV/r) as a comparator contributed data

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service