Skip to Content
Merck
All Photos(1)

Key Documents

SML2091

Sigma-Aldrich

Dalcetrapib

≥97% (HPLC)

Synonym(s):

JTT-705, RO-4607381, S-[2-[[1-(2-ethylbutyl)cyclohexane]carbonylamino]phenyl]-2-methylpropanethioate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$172.00
25 MG
$632.00

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$172.00
25 MG
$632.00

About This Item

Empirical Formula (Hill Notation):
C23H35NO2S
CAS Number:
Molecular Weight:
389.59
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$172.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥97% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(C)C(SC1=CC=CC=C1NC(C2(CCCCC2)CC(CC)CC)=O)=O

InChI

1S/C23H35NO2S/c1-5-18(6-2)16-23(14-10-7-11-15-23)22(26)24-19-12-8-9-13-20(19)27-21(25)17(3)4/h8-9,12-13,17-18H,5-7,10-11,14-16H2,1-4H3,(H,24,26)

InChI key

YZQLWPMZQVHJED-UHFFFAOYSA-N

Biochem/physiol Actions

Cholesteryl ester transfer protein (CETP) inhibitor; increases serum levels of HDL-cholesterol
Dalcetrapib is a cholesteryl ester transfer protein (CETP) inhibitor. It increases HDL levels, but showed variable results on clinical outcomes. Clinical outcomes, effects on C-reactive protein and cholesterol efflux have been found to be genotype-dependent, influenced by correlated polymorphisms in the ADCY9 (adenylate cyclase type 9) gene. Positive results were obtained with AA genotype of ADCY9 gene whereas the GG genotype showed negative results and the AG genotype was neutral.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marie-Jeanne Bertrand et al.
Expert opinion on emerging drugs, 22(1), 1-26 (2016-12-09)
Cardiovascular (CV) atherosclerotic disease remains the leading cause of morbidity and mortality worldwide, despite the advances in contemporary therapies. Inflammation is an important process in atherosclerosis, leading to plaque rupture and acute coronary syndrome. Although statin therapy has substantially reduced
Jean-Claude Tardif et al.
Arteriosclerosis, thrombosis, and vascular biology, 37(3), 396-400 (2017-01-28)
High-density lipoproteins are involved in reverse cholesterol transport and possess anti-inflammatory and antioxidative properties. Paradoxically, CETP (cholesteryl ester transfer protein) inhibitors have been shown to increase inflammation as revealed by a raised plasma level of high-sensitivity C-reactive protein. CETP inhibitors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service