Skip to Content
Merck
All Photos(1)

Documents

SML2083

Sigma-Aldrich

[Ala13]-Apelin-13 trifluoroacetate salt

≥96% (HPLC)

Synonym(s):

(F13A)apelin-13, Alanine apelin-13, Apelin-13(F13A), Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Ala TFA, L-glutaminyl-L-arginyl-L-prolyl-L-arginyl-L-leucyl-L-seryl-L-histidyl-L-lysylglycyl-L-prolyl-L-methionyl-L-prolyl-L-alanine TFA, QRPRLSHKGPMPA TFA, [Ala-13] apelin-13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H107N23O16S · xC2HF3O2
CAS Number:
Molecular Weight:
1474.73 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥96% (HPLC)

form

powder

color

white to beige

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

(Ala13)-Apelin-13 corresponds to Apelin-13 with a Phe-to-Ala substitution at the C-terminus (F13A) and functions as an antagonist against apelin-dependent APJ functions both in cultures and in animal studies in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

İlknur Birsen et al.
The journal of physiological sciences : JPS, 67(3), 373-385 (2016-07-03)
The objective of this study was to explore the role of apelin in the healing of gastric lesions induced by stress. Male Wistar rats were exposed to water immersion and restraint stress (WIRS) for 6 h with or without the apelin
Xue Han et al.
Atherosclerosis, 244, 1-8 (2015-11-19)
Vascular calcification (VC) is closely related to cardiovascular events in chronic kidney disease (CKD). Apelin has emerged as a potent regulator of cardiovascular function, but its role in VC during CKD remains unknown. We determined whether apelin plays a role
Dennis K Lee et al.
Endocrinology, 146(1), 231-236 (2004-10-16)
The apelin peptide is the endogenous ligand for the apelin G protein-coupled receptor. The distribution of the apelin peptides and receptor are widespread in the central nervous system and periphery, with reported roles in the hypothalamic-pituitary-adrenal axis, blood pressure regulation
Mehmet Bülbül et al.
American journal of physiology. Gastrointestinal and liver physiology, 310(4), G249-G261 (2015-12-19)
Apelin, an endogenous ligand for APJ receptor, has been reported to be upregulated in paraventricular nucleus (PVN) following stress. Central apelin is known to stimulate release of corticotropin-releasing factor (CRF) via APJ receptor. We tested the hypothesis that stress-induced gastrointestinal
Anne Drougard et al.
Antioxidants & redox signaling, 20(4), 557-573 (2013-07-25)
We have previously demonstrated that central apelin is implicated in the control of peripheral glycemia, and its action depends on nutritional (fast versus fed) and physiological (normal versus diabetic) states. An intracerebroventricular (icv) injection of a high dose of apelin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service