Skip to Content
Merck
All Photos(1)

Key Documents

SML2017

Sigma-Aldrich

Compound C108

≥98% (HPLC)

Synonym(s):

2-Hydroxy-2-[1-(2-hydroxyphenyl)ethylidene]hydrazide-benzoic acid, 2-Hydroxy-N′-[1-(2-hydroxyphenyl)ethylidene]benzohydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$424.00

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$132.00
25 MG
$424.00

About This Item

Empirical Formula (Hill Notation):
C15H14N2O3
CAS Number:
Molecular Weight:
270.28
UNSPSC Code:
12352200
NACRES:
NA.77

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

ambient

storage temp.

−20°C

SMILES string

OC1=C(C=CC=C1)C(C)=NNC(C2=C(C=CC=C2)O)=O

Biochem/physiol Actions

C108 is a small molecule that exhibits cancer-selective cytotoxicity (Viability = 100%/MCF-10A vs. 50%/BT-474, 67%/4T1, 70%/MDA-MB-231 & MDA-MB-453 post 48 h 1 μM C108 treatment) and synergizes with low dose paclitaxel in reducing ALDH-positive tumor-initiating cells (TIC poulation = 56.6%/control, 60.6%/0.1 μM paclitaxel alone, 47.4%/1 μM C108 alone, 7.3%/combined treatment for 24 h in BT474 breast cancer cultures) by targeting stress granule-associated protein G3BP2 (GAP SH3 domain-binding protein 2). Likewise, C108 pretreatment prior to xenografting greatly reduces BT-474 tumor-initiating frequency (from 1/175 to 1/1103 by limiting dilution xenograft assays) in mice in vivo. G3BP2 is reported to bind and stabilize SART3 mRNA, thereby indirectly regulating the core pluripotency transcription factors Oct-4 and Nanog critically involved in ESC self-renewal and breast tumor initiation.
G3BP2 inhibitor that exhibits cancer-selective toxicity and synergizes with low dose paclitaxel against tumor-initiating cell (TIC) population in breast cancer cultures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nisha Gupta et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(5), 1033-1038 (2017-01-18)
Breast tumors contain tumorigenic cancer cells, termed "tumor-initiating cells" (TICs), which are capable of both replenishing themselves and giving rise to populations of nontumorigenic breast cancer cells (non-TICs). However, the molecular mechanisms responsible for breast tumor initiation remain poorly understood.
Ningning Li et al.
Current pharmaceutical biotechnology, 21(11), 1107-1118 (2020-03-21)
Renal fibrosis is a common pathway leading to the progression of chronic kidney disease. Activated fibroblasts contribute remarkably to the development of renal fibrosis. Although apigenin has been demonstrated to play a protective role from fibrotic diseases, its pharmacological effect

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service