Skip to Content
Merck
All Photos(1)

Key Documents

SML1957

Sigma-Aldrich

ML RR-S2 CDA sodium salt

≥95% (HPLC)

Synonym(s):

Dithio-(Rp, Rp)-2′,3′-CDA sodium salt, Dithio-(Rp, Rp)-2′,3′-c-diAMP sodium salt, Dithio-(Rp, Rp)-2′,5′-3′,5′-c-diAMP sodium salt, Dithio-(Rp, Rp)-[cyclic[A(2′,5′)pA(3′,5′)p]] sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

73.5 μG
$417.00

$417.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
73.5 μG
$417.00

About This Item

Empirical Formula (Hill Notation):
C20H24N10O10P2S2 · xNa+
CAS Number:
Molecular Weight:
690.54 (free acid basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$417.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

shipped in

wet ice

storage temp.

−20°C

SMILES string

NC1=NC=NC2=C1N=CN2[C@H]3[C@H](O)[C@H](OP4([S-])=O)[C@@H](COP(O[C@H]5[C@@H](O)[C@H](N(C=N6)C7=C6C(N)=NC=N7)O[C@@H]5CO4)([S-])=O)O3.[R].[R].[Na+]

Biochem/physiol Actions

A mixed linkage (ML) cyclic di-adenosine bisthiophosphate with potent STING-activating activity and in vivo anti-tumor efficacy.
ML-RR-S2 CDA is a potent STING-activating cyclic di-adenosine (CDA or c-diAMP) nucleotide ligand with the 2′-5′, 3′-5′ mixed phosphodiester linkage (ML) in the [Rp,Rp] configuration and doubly substituted thiophosphate (S2) for optimal phosphodiesterase-resistance and thereby enhanced cellular potency. ML-RR-S2 CDA effectively induces IRF3, NF-kB, and STAT6 transcription activities in a STING-dependent manner in both murine (DC2.4 & BMMs) and human (THP1 & PBMCs) cultures (5-100 μM) as well as displays profound anti-tumor efficacy in vivo (100% growth inhibition of established B16.F10, 4T-1 or CT26 tumor with 3X 50 μg/mouse/96 hr intratumoral injections) with lasting immune-mediated tumor rejection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily Curran et al.
Cell reports, 15(11), 2357-2366 (2016-06-07)
Type I interferon (IFN), essential for spontaneous T cell priming against solid tumors, is generated through recognition of tumor DNA by STING. Interestingly, we observe that type I IFN is not elicited in animals with disseminated acute myeloid leukemia (AML). Further
Leticia Corrales et al.
Cell reports, 11(7), 1018-1030 (2015-05-12)
Spontaneous tumor-initiated T cell priming is dependent on IFN-β production by tumor-resident dendritic cells. On the basis of recent observations indicating that IFN-β expression was dependent upon activation of the host STING pathway, we hypothesized that direct engagement of STING
Juan Fu et al.
Science translational medicine, 7(283), 283ra52-283ra52 (2015-04-17)
Stimulator of interferon genes (STING) is a cytosolic receptor that senses both exogenous and endogenous cytosolic cyclic dinucleotides (CDNs), activating TBK1/IRF3 (interferon regulatory factor 3), NF-κB (nuclear factor κB), and STAT6 (signal transducer and activator of transcription 6) signaling pathways

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service