Skip to Content
Merck
All Photos(1)

Key Documents

SML1952

Sigma-Aldrich

GAT228

≥98% (HPLC)

Synonym(s):

3-[(1R)-2-nitro-1-phenylethyl]-2-phenyl-1H-Indole, R-(+)-3-(2-Nitro-1-phenylethyl)-2-phenyl-1H-indole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$126.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$126.00

About This Item

Empirical Formula (Hill Notation):
C22H18N2O2
CAS Number:
Molecular Weight:
342.39
UNSPSC Code:
12352200
NACRES:
NA.77

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 80 to 94°, c = 1 in methanol

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=[N+]([O-])C[C@H](C1=CC=CC=C1)C2=C(C3=CC=CC=C3)NC4=CC=CC=C42

InChI

1S/C22H18N2O2/c25-24(26)15-19(16-9-3-1-4-10-16)21-18-13-7-8-14-20(18)23-22(21)17-11-5-2-6-12-17/h1-14,19,23H,15H2/t19-/m1/s1

InChI key

OHZDCJJHWPHZJD-LJQANCHMSA-N

Biochem/physiol Actions

GAT228 is the R-(+)-enantiomer of GAT211, a positive allosteric modulator (PAM) of cannabinoid CB1 receptor signaling that was found to amplify the therapeutic effect of endocannabinoids without the negative side effects of psychoactivity or tolerance. GAT228 was found to be an unbiased CB1 allosteric agonist, while the S-(-)-enantiomer, GAT229,was found to be a potent, Gαi/o-biased CB1 PAM without intrinsic activity. In radioligand binding assays, both GAT228 and GAT229 behaved as PAMs of orthosteric ligand binding. Allosteric CB1R activation by GAT211 and its enantiomers could be a better therapeutic strategy for enhancing endogenous cannabinergic activity than targeting endocannabinoid-degrading enzymes with small-molecule inhibitors, with a lower likelihood of tolerance and dependence.
Unbiased cannabinoid CB1 allosteric agonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert B Laprairie et al.
ACS chemical neuroscience, 8(6), 1188-1203 (2017-01-20)
The cannabinoid 1 receptor (CB1R) is one of the most widely expressed metabotropic G protein-coupled receptors in brain, and its participation in various (patho)physiological processes has made CB1R activation a viable therapeutic modality. Adverse psychotropic effects limit the clinical utility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service