Skip to Content
Merck
All Photos(1)

Key Documents

SML1940

Sigma-Aldrich

ADDA 5 hydrochloride

≥98% (HPLC)

Synonym(s):

1-[2-(1-Adamantyl)ethoxy]-3-(3,4-dihydro-2(1H)-isoquinolinyl)-2-propanol hydrochloride, 3,4-Dihydro-α-[(2-tricyclo[3.3.1.13,7]dec-1-ylethoxy)methyl]-2(1H)-isoquinolineethanol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$132.00
25 MG
$424.00

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$132.00
25 MG
$424.00

About This Item

Empirical Formula (Hill Notation):
C24H35NO2 · HCl
CAS Number:
Molecular Weight:
406.00
UNSPSC Code:
12352200
NACRES:
NA.77

$132.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CN1CCC(C=CC=C2)=C2C1)COCCC34C[C@H]5C[C@H](C[C@H](C5)C4)C3.[H]Cl

Application

ADDA 5 hydrochloride has been used as a complex IV inhibitor to study its effects on leptomeningeal derivatives of human breast and lung cancer cells.[1]

Biochem/physiol Actions

ADDA 5 is a potent and selective inhibitor of cytochrome c oxidase (Complex IV) that depletes the bioenergetics reserve capacity in intact glioma cells. ADDA 5 specifically inhibits the proliferation of chemosensitive and chemoresistant glioma cells, while sparing non-cancer cells. Additionally ADDA 5 is effective against glioma stem cells.
Potent and selective inhibitor of cytochrome c oxidase (Complex IV)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Remsik et al.
Cancer reports (Hoboken, N.J.), e1236-e1236 (2020-12-30)
Leptomeningeal metastasis (LM), or spread of cancer cells into the cerebrospinal fluid (CSF), is characterized by a rapid onset of debilitating neurological symptoms and markedly bleak prognosis. The lack of reproducible in vitro and in vivo models has prevented the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service