Skip to Content
Merck
All Photos(1)

Key Documents

SML1935

Sigma-Aldrich

Nimorazole

≥98% (HPLC)

Synonym(s):

4-(2-(5-Nitro-1H-imidazol-1-yl)ethyl)morpholine, K 1900, K-1900, K1900, NSC 107524, NSC-107524, NSC107524

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$48.50
50 MG
$164.00

$48.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$48.50
50 MG
$164.00

About This Item

Empirical Formula (Hill Notation):
C9H14N4O3
CAS Number:
Molecular Weight:
226.23
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

$48.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

[N+](=O)([O-])c1[n](cnc1)CCN2CCOCC2

InChI

1S/C9H14N4O3/c14-13(15)9-7-10-8-12(9)2-1-11-3-5-16-6-4-11/h7-8H,1-6H2

InChI key

MDJFHRLTPRPZLY-UHFFFAOYSA-N

Biochem/physiol Actions

A nitroimidazole-based hypoxic cell-radiation sensitizer and anti-infective against Trichomonas vaginalis.
Nimorazole is a nitroimidazole-based hypoxic cell-radiation sensitizer with less toxicity than etanidazole or misonidazole. Under low oxygen (hypoxic) condition, reductive activation of its 5-nitroimidazole moiety leads to enhanced adduct formation with reduced glutathione, which form the basis of cancer/tumour-selective radiation sensitization. Nimorazole is also a known anti-infective agent against trichomoniasis, an infectious disease caused by the protozoan parasite Trichomonas vaginalis.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 1B - Repr. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas R Wittenborn et al.
Acta oncologica (Stockholm, Sweden), 54(9), 1385-1392 (2015-09-04)
Hypoxia is a characteristic feature of solid tumours that significantly reduces the efficacy of conventional radiation therapy. In this study we investigated the role of hypoxia in a stereotactic radiation schedule by using a variety of hypoxic modifiers in a
Thomas M Ashton et al.
Nature communications, 7, 12308-12308 (2016-07-28)
Tumour hypoxia renders cancer cells resistant to cancer therapy, resulting in markedly worse clinical outcomes. To find clinical candidate compounds that reduce hypoxia in tumours, we conduct a high-throughput screen for oxygen consumption rate (OCR) reduction and identify a number
Radiotherapy quality assurance of the IAEA-HypoX trial of the accelerated radiotherapy in the treatment of head and neck squamous cell carcinoma with or without the hypoxic radiosensitizer nimorazole.
Mohamed A Hassan Metwally et al.
Acta oncologica (Stockholm, Sweden), 54(9), 1673-1677 (2015-09-24)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service