Skip to Content
Merck

Skip To

SML1925

Bepristat 2a hydrochloride

≥95% (HPLC)

Synonym(s):

6-Bromo-2-[[[2-(diethylamino)ethyl]amino]methyl]-5-hydroxy-1-phenyl-1H-indole-3-carboxylic acid ethyl ester hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mg
Check Cart for Availability
$153.00
25 mg
Check Cart for Availability
$620.00

About This Item

Empirical Formula (Hill Notation):
C24H30BrN3O3 · xHCl
CAS Number:
Molecular Weight:
488.42 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCN(CC)CCNCC1=C(C(OCC)=O)C2=CC(O)=C(Br)C=C2N1C3=CC=CC=C3.[H]Cl

InChI key

HKOKWSVFJJFYDV-UHFFFAOYSA-N

Biochem/physiol Actions

Bepristat 2a is a selective reversible inhibitor of protein disulfide isomerase (PDI), an enzyme in the endoplasmic reticulum that catalyzes disulfide bond breakage and reformation to catalyze protein folding. Unlike most PDI inhibitors, Bepristat 2a binds at the substrate-binding site, rather than the catalytic site. Bepristat 2a blocked PDI activity with an IC50 value of 1200 nM, while enhancing catalytic activity of remote PDI domains. PDI is up-regulated in several cancers, has been implicated in neurodegenerative processes, and plays an important role in thrombus formation. Bepristat 2a potently inhibited platelet aggregation and thrombus formation in vivo.
Bepristat 2a is a selective reversible inhibitor of protein disulfide isomerase (PDI).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L1011SML0860SML0853
description

≥95% (HPLC)

description

>98% (TLC), powder

description

≥98% (HPLC)

description

≥98% (HPLC)

assay

≥95% (HPLC)

assay

>98% (TLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

powder

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage temp.

2-8°C

solubility

H2O: 10 mg/mL, clear

solubility

-

solubility

DMSO: 20 mg/mL, clear

solubility

H2O: 1 mg/mL, clear (warmed)

storage condition

desiccated

storage condition

-

storage condition

desiccated

storage condition

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters