Skip to Content
Merck
All Photos(1)

Key Documents

SML1907

Sigma-Aldrich

ESI-05

≥98% (HPLC)

Synonym(s):

1,3,5-Trimethyl-2-[(4-methylphenyl)sulfonyl]-benzene, Mesityl (4-methylphenyl) sulfone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$452.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$452.00

About This Item

Empirical Formula (Hill Notation):
C16H18O2S
CAS Number:
Molecular Weight:
274.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC1=CC=C(S(C2=C(C)C=C(C)C=C2C)(=O)=O)C=C1

InChI

1S/C16H18O2S/c1-11-5-7-15(8-6-11)19(17,18)16-13(3)9-12(2)10-14(16)4/h5-10H,1-4H3

InChI key

CGPHOZWFSFNOEQ-UHFFFAOYSA-N

Application

ESI-05 has been used to study the effect of protein exchange directly activated by cAMP 2 on traumatic brain injury.[1]

Biochem/physiol Actions

ESI-05 is a selective inhibitor against cAMP-regulated guanine nucleotide exhange factor (GEF) EPAC2 (Exchange factor directly activated by cAMP 2) activation by targeting an EPAC2-specific allosteric site at the interface of the two cAMP-binding domains.
ESI-05 is a selective inhibitor against cAMP-regulated guanine nucleotide exhange factor (GEF) EPAC2 (Exchange factor directly activated by cAMP 2) activation by targeting an EPAC2-specific allosteric site at the interface of the two cAMP-binding domains. ESI-05 inhibits EAPC2, but not EPAC1 or PKA, activation by cAMP in a highly potent (IC50 = 430 nM; 25 μM cAMP) and cAMP-competitive manner in cell-free assays, and suppresses cAMP analog 007-AM-induced cellular Rap1 activation in EPAC2-, but not EPAC1-, expressing HEK293 transfectants.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Calderón-Sánchez et al.
PloS one, 11(2), e0147375-e0147375 (2016-02-04)
Urocortin-1 (Ucn-1) is an endogenous peptide that protects heart from ischemia and reperfusion (I/R) injuries. Ucn-1 is known to prevent cardiac cell death, but its role in the transcription of specific genes related to survival signaling pathway has not been
Tamara Tsalkova et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(45), 18613-18618 (2012-10-24)
The major physiological effects of cAMP in mammalian cells are transduced by two ubiquitously expressed intracellular cAMP receptors, protein kinase A (PKA) and exchange protein directly activated by cAMP (EPAC), as well as cyclic nucleotide-gated ion channels in certain tissues.
Haijun Chen et al.
Journal of medicinal chemistry, 56(3), 952-962 (2013-01-05)
EPAC1 and EPAC2, two isoforms of exchange proteins directly activated by cAMP (EPAC), respond to the second messenger cAMP and regulate a wide variety of intracellular processes under physiological and pathophysiological circumstances. Herein, we report the chemical design, synthesis, and
Ling Zhang et al.
Frontiers in neuroscience, 12, 263-263 (2018-05-10)
Traumatic brain injury (TBI) is a major cause of mortality and disability worldwide. TBI-induced neuronal apoptosis is one of the main contributors to the secondary injury process. The aim of this study is to investigate the involvement of Exchange protein

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service