Skip to Content
Merck
All Photos(1)

Documents

SML1892

Sigma-Aldrich

JPH203 hydrochloride

≥98% (HPLC)

Synonym(s):

(S)-2-Amino-3-(4-((5-amino-2-phenylbenzo[d]oxazol-7-yl)methoxy)-3,5-dichlorophenyl)propanoic acid hydrochloride, JPH 203 hydrochloride, KYT 0353 hydrochloride, KYT-0353 hydrochloride, KYT0353 hydrochloride, O-(5-Amino-2-phenylbenzoxazole-7-yl)methyl-3,5-dichloro-L-tyrosine hydrochloride, O-[(5-Amino-2-phenyl-7-benzoxazolyl)methyl]-3,5-dichloro-L-tyrosine dihydrochloride, SCHEMBL17360639 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H19Cl2N3O4 · xHCl
CAS Number:
Molecular Weight:
472.32 (free base basis)
MDL number:
UNSPSC Code:
12352200

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(O)[C@@H](N)CC1=CC(Cl)=C(OCC2=CC(N)=CC3=C2OC(C4=CC=CC=C4)=N3)C(Cl)=C1

Biochem/physiol Actions

JPH203 (KYT-0353) selectively inhibits L-type amino acid transporter LAT1-dependent L-leucine uptake and growth in HT-29 and human LAT1-expressing S2 murine renal proximal tubule cell cultures, but not human LAT2-expressing S2 cells (IC50/GI50 = 0.06/4.1, 0.14/16.4, and >10/>1000 μM, respectively). When administered via intravenous injection (6.3-25 mg/kg/d from d0 to d13), JPH203 effectively inhibits HT-29-derived tumor growth in mice in vivo.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koji Oda et al.
Cancer science, 101(1), 173-179 (2009-11-11)
Most tumor cell membranes overexpress L-type amino acid transporter 1, while normal cell membranes contain l-type amino acid transporter 2; both are Na(+)-independent amino acid transporters. Therefore, compounds that selectively inhibit L-type amino acid transporter 1 offer researchers with a
Michael F Wempe et al.
Drug metabolism and pharmacokinetics, 27(1), 155-161 (2011-09-15)
Many primary human tumors and tumor cell lines highly express human L-type amino acid transporter 1 (hLAT1); cancerous cells in vivo are strongly linked to LAT1 expression. Synthetic chemistry and in vitro screening efforts have afforded a variety of novel
Gyujin Park et al.
JCI insight, 8(7) (2023-03-03)
Hypothalamic neurons regulate body homeostasis by sensing and integrating changes in the levels of key hormones and primary nutrients (amino acids, glucose, and lipids). However, the molecular mechanisms that enable hypothalamic neurons to detect primary nutrients remain elusive. Here, we
Junko Toyoshima et al.
Journal of pharmaceutical sciences, 102(9), 3228-3238 (2013-05-29)
JPH203 has been developed as an anticancer drug that inhibits L-type amino acid transporter 1-mediated essential amino acid uptake into tumor cells. This study sought to elucidate which drug transporters may be involved in JPH203 hepatic elimination, and to estimate
Yi-Heng Liu et al.
International journal of molecular sciences, 22(20) (2021-10-24)
The l-type amino acid transporter 1 (LAT1) is a membranous transporter that transports neutral amino acids for cells and is dysregulated in various types of cancer. Here, we first observed increased LAT1 expression in pemetrexed-resistant non-small cell lung cancer (NSCLC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service