Skip to Content
Merck
All Photos(1)

Key Documents

SML1884

Sigma-Aldrich

Mitoglitazone

≥98% (HPLC)

Synonym(s):

5-[[4-[2-(5-ethyl-2-pyridinyl)-2-oxoethoxy]phenyl]methyl]-2,4-thiazolidinedione, MSDC-0160

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$360.00

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C19H18N2O4S
CAS Number:
Molecular Weight:
370.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CCC1=CN=C(C(COC2=CC=C(CC3SC(NC3=O)=O)C=C2)=O)C=C1

InChI

1S/C19H18N2O4S/c1-2-12-5-8-15(20-10-12)16(22)11-25-14-6-3-13(4-7-14)9-17-18(23)21-19(24)26-17/h3-8,10,17H,2,9,11H2,1H3,(H,21,23,24)

InChI key

IRNJSRAGRIZIHD-UHFFFAOYSA-N

Biochem/physiol Actions

MSDC-0160 is an mTOT (mitochondrial target of thiazolidinediones) modulator that targets the mitochondrial pyruvate carrier (MPC), which is a key controller of cellular metabolism. MSDC-0160 is reported to enhance the cells′ ability to handle potentially harmful proteins, which leads to reduced inflammation and less nerve cell death. It has already been in clinical trials for diabetes and Alzheimer′s disease and clinical trials for Parkinson′s Disease are planned.
Mitochondrial pyruvate carrier (MPC) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raj C Shah et al.
Current Alzheimer research, 11(6), 564-573 (2014-06-17)
Alzheimer's disease (AD) is associated with insulin resistance and specific regional declines in cerebral metabolism. The effects of a novel mTOT modulating insulin sensitizer (MSDC-0160) were explored in non-diabetic patients with mild AD to determine whether treatment would impact glucose
Nidhi Rohatgi et al.
PloS one, 8(5), e62012-e62012 (2013-05-08)
Major bottlenecks in the expansion of human β-cell mass are limited proliferation, loss of β-cell phenotype, and increased apoptosis. In our previous studies, activation of Wnt and mTOR signaling significantly enhanced human β-cell proliferation. However, isolated human islets displayed insulin
Anamitra Ghosh et al.
Science translational medicine, 8(368), 368ra174-368ra174 (2016-12-09)
Mitochondrial and autophagic dysfunction as well as neuroinflammation are involved in the pathophysiology of Parkinson's disease (PD). We hypothesized that targeting the mitochondrial pyruvate carrier (MPC), a key controller of cellular metabolism that influences mTOR (mammalian target of rapamycin) activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service