Skip to Content
Merck
All Photos(1)

Key Documents

SML1876

Sigma-Aldrich

PaPE-1

≥98% (HPLC)

Synonym(s):

(S)-5-(4-Hydroxy-3,5-dimethyl-phenyl)-indan-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$164.00

$164.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$164.00

About This Item

Empirical Formula (Hill Notation):
C17H18O2
CAS Number:
Molecular Weight:
254.32
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$164.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O[C@H]1CCC2=C1C=CC(C3=CC(C)=C(O)C(C)=C3)=C2

InChI

1S/C17H18O2/c1-10-7-14(8-11(2)17(10)19)12-3-5-15-13(9-12)4-6-16(15)18/h3,5,7-9,16,18-19H,4,6H2,1-2H3/t16-/m0/s1

InChI key

UCXWFKNKSWDWCD-INIZCTEOSA-N

Biochem/physiol Actions

PaPE-1 is a "Pathway Preferential Estrogen" that activates the extranuclear signaling pathway without activating the nuclear signaling pathway. PaPE-1 bound 50,000-fold less well to ERα and Erβ estrogen receptors. PaPE-1 activated extranuclear-initiated ER-regulated genes, but showed essentially no activation of nuclear-initiated estrogen receptor (ER) gene targets such as the progesterone receptor. Unlike estradiol (E2), PaPE-1 did not stimulate proliferation of MCF-7 breast cancer cells. Like estradiol, PaPE-1 strongly activated MAPK and mTOR pathway. Instead, it showed preferential estrogen-like activity in non-reproductive (metabolic and vascular) tissues, reducing body weight gain and fat accumulation in ovariectomized mice and accelerating repair of endothelial damage in the vascualture.
PaPE-1 is a "Pathway Preferential Estrogen" that activates the extranuclear signaling pathway without activating the nuclear signaling pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeynep Madak-Erdogan et al.
Science signaling, 9(429), ra53-ra53 (2016-05-26)
There is great medical need for estrogens with favorable pharmacological profiles that support desirable activities for menopausal women, such as metabolic and vascular protection, but that lack stimulatory activities on the breast and uterus. We report the development of structurally
Laura Marroqui et al.
Chemosphere, 265, 129051-129051 (2020-12-01)
Bisphenol-S (BPS) and Bisphenol-F (BPF) are current Bisphenol-A (BPA) substitutes. Here we used pancreatic β-cells from wild type (WT) and estrogen receptor β (ERβ) knockout (BERKO) mice to investigate the effects of BPS and BPF on insulin secretion, and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service