Skip to Content
Merck
All Photos(1)

Key Documents

SML1866

Sigma-Aldrich

KIN1408

≥98% (HPLC)

Synonym(s):

7-((4-(Difluoromethoxy)phenyl)(5-methoxybenzo[d]thiazol-2-ylamino)methyl)quinolin-8-ol, 7-[[4-(Difluoromethoxy)phenyl][(5-methoxy-2-benzothiazolyl)amino]methyl]-8-quinolinol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$452.00

$112.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$452.00

About This Item

Empirical Formula (Hill Notation):
C25H19F2N3O3S
CAS Number:
Molecular Weight:
479.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 30 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(C(N=CC=C1)=C1C=C2)=C2C(C3=CC=C(OC(F)F)C=C3)NC4=NC5=C(S4)C=CC(OC)=C5

InChI key

YSGBFDHVEQJPPX-UHFFFAOYSA-N

Biochem/physiol Actions

KIN1408 is a small molecule that targets factors at or above the level of MAVS in the RLR signaling pathway to drive IRF3 activation (IRF3 nuclear translocation ECmax = 5 μM in 20 h; Huh7 cells) without significant cytotoxicity (50 μM/20 h in Huh7 or 20 μM/36 h in HEK293 cultures). KIN1408 induces cellular transcription of innate immune genes (Eff. conc. 1.25-20 μM in 20 h; PMA-differentiated THP-1 cells) in a MAVS- and IRF3-dependent manner and exhibits broad-spectrum anti-viral activity (Eff. conc. 1-5 μM), including dengue virus 2 (DV2; Huh7), influenza A (IAV H3N2; HEK293), RSV (A2 strain; HeLa), Ebola (EBOV strain Zaire; HUVECs), Nipah virus (NiV; HUVECs), and Lassa virus (LASV; HUVECs). MAVS signaling activation upon Zika virus infection or KIN1408 treatment is reported to result in pTBK1 mitochondria relocalization and caspase 3-mediated apoptosis in human neuroepithelial stem (NES) cells.
KIN1408 is a small molecule that targets factors at or above the level of MAVS in the RLR signaling pathway.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Onorati et al.
Cell reports, 16(10), 2576-2592 (2016-08-29)
The mechanisms underlying Zika virus (ZIKV)-related microcephaly and other neurodevelopment defects remain poorly understood. Here, we describe the derivation and characterization, including single-cell RNA-seq, of neocortical and spinal cord neuroepithelial stem (NES) cells to model early human neurodevelopment and ZIKV-related
R R Green et al.
Genomics data, 7, 290-292 (2016-03-17)
The recognition of pathogen associated molecular patterns (PAMPs) by pattern recognition receptors (PRR) during viral infection initiates the induction of antiviral signaling pathways, including activation of the Interferon Regulator Factor 3 (IRF3). We identified small molecule compounds that activate IRF3
Sowmya Pattabhi et al.
Journal of virology, 90(5), 2372-2387 (2015-12-18)
The cellular response to virus infection is initiated when pathogen recognition receptors (PRR) engage viral pathogen-associated molecular patterns (PAMPs). This process results in induction of downstream signaling pathways that activate the transcription factor interferon regulatory factor 3 (IRF3). IRF3 plays

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service