Skip to Content
Merck
All Photos(1)

Key Documents

SML1864

Sigma-Aldrich

OUL35

≥98% (HPLC)

Synonym(s):

4,4′-Oxybis-benzamide, 4-(4-Carbamoylphenoxyl)benzamide, NSC 39047, NSC39047

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$452.00

$112.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$452.00

About This Item

Empirical Formula (Hill Notation):
C14H12N2O3
CAS Number:
Molecular Weight:
256.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$112.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N)C1=CC=C(OC2=CC=C(C(N)=O)C=C2)C=C1

InChI

1S/C14H12N2O3/c15-13(17)9-1-5-11(6-2-9)19-12-7-3-10(4-8-12)14(16)18/h1-8H,(H2,15,17)(H2,16,18)

InChI key

XZRCQWLPMXFGHE-UHFFFAOYSA-N

Biochem/physiol Actions

OUL35 is a cell penetrant, potent and selective ARTD10/PARP10 inhibitor.
OUL35 is a cell penetrant, potent and selective ARTD10/PARP10 inhibitor. OUL35 rescues HeLa cells from ARTD10 induced apoptotic cell death.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service