Skip to Content
Merck
All Photos(1)

Key Documents

SML1798

Sigma-Aldrich

10-Cl-BBQ

≥98% (HPLC)

Synonym(s):

10-Chloro-7H-benzo[de]benzo[4,5]imidazo[2,1-a]isoquinolin-7-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$143.00
25 MG
$583.00

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
5 MG
$143.00
25 MG
$583.00

About This Item

Empirical Formula (Hill Notation):
C18H9ClN2O
CAS Number:
Molecular Weight:
304.73
UNSPSC Code:
12352200
NACRES:
NA.77

$143.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 0.5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

O=C(C1=C2C(C=CC=C23)=CC=C1)N4C3=NC5=C4C=C(Cl)C=C5

InChI

1S/C18H9ClN2O/c19-11-7-8-14-15(9-11)21-17(20-14)12-5-1-3-10-4-2-6-13(16(10)12)18(21)22/h1-9H

InChI key

ZJESJVGSDQOZAT-UHFFFAOYSA-N

Biochem/physiol Actions

10-Cl-BBQ is an orally bioavailable, non-toxic benzimidazoisoquinoline derivative that acts as an aryl hydrocarbon receptor (AhR) agonist via directly binding to AhR (IC50  = 2.6 nM in a competitive binding assay) and induces its nuclear translocation. 10-Cl-BBQ is shown to increase CD4+  T-cells that co-express CD25, CTLA-4 and ICOS, as well as several other genes associated with regulatory T cell (Treg) function in a graft versus host response model (GVH, C57BI/6 T cells injected into B6D2F1 host mice). 10-Cl-BBQ displays good pharmacokinetic profile in mice (t1/2 = 2 h, Cmax = 21.5 μg/L, 10 mg/kg, i.p.). Also shown to prevent insulitis in a NOD T1D mouse model which is independent of Fox3 +  Tregs (60 mg/kg, p.o, q.o.d.).
10-Cl-BBQ is an orally bioavailable, non-toxic benzimidazoisoquinoline derivative.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sumit Punj et al.
PloS one, 9(2), e88726-e88726 (2014-03-04)
The aryl hydrocarbon receptor (AhR) is a ligand-activated transcription factor that plays multiple roles in regulation of immune and inflammatory responses. The ability of certain AhR ligands to induce regulatory T cells (Tregs) has generated interest in developing AhR ligands
Allison K Ehrlich et al.
Journal of immunology (Baltimore, Md. : 1950), 196(1), 264-273 (2015-11-18)
Aryl hydrocarbon receptor (AhR) activation by high-affinity ligands mediates immunosuppression in association with increased regulatory T cells (Tregs), making this transcription factor an attractive therapeutic target for autoimmune diseases. We recently discovered 10-chloro-7H-benzimidazo[2,1-a]benzo[de]iso-quinolin-7-one (10-Cl-BBQ), a nanomolar affinity AhR ligand with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service