Skip to Content
Merck
All Photos(1)

Key Documents

SML1765

Sigma-Aldrich

DPBQ

≥98% (HPLC)

Synonym(s):

2,3-Diphenylbenzo[g]quinoxaline-5,10-dione, NSC 629713

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H14N2O2
CAS Number:
Molecular Weight:
362.38
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

, light green to dark green

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C(N=C(C3=CC=CC=C3)C(C4=CC=CC=C4)=N2)C(C5=CC=CC=C51)=O

Biochem/physiol Actions

DPBQ is a potent apoptosis inducer that is specific for high-ploidy cells. It appears that DPBQ induces expression and phosphorylation of p53 and this effect is specific to tetraploid cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service