Skip to Content
Merck
All Photos(1)

Key Documents

SML1761

Sigma-Aldrich

AA-CW236

≥98% (HPLC)

Synonym(s):

4-(2-(5-(Chloromethyl)-4-(4-(trifluoromethoxy)phenyl)-1H-1,2,3-triazol-1-yl)ethyl)-3,5-dimethylisoxazole

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$496.00

$496.00


Estimated to ship onMay 29, 2025



Select a Size

Change View
25 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C17H16ClF3N4O2
CAS Number:
Molecular Weight:
400.78
UNSPSC Code:
12352200
NACRES:
NA.77

$496.00


Estimated to ship onMay 29, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL protein, clear

storage temp.

2-8°C

SMILES string

CC1=C(CCN2N=NC(C3=CC=C(OC(F)(F)F)C=C3)=C2CCl)C(C)=NO1

InChI

1S/C17H16ClF3N4O2/c1-10-14(11(2)27-23-10)7-8-25-15(9-18)16(22-24-25)12-3-5-13(6-4-12)26-17(19,20)21/h3-6H,7-9H2,1-2H3

InChI key

MPDMMSRZEHOFPA-UHFFFAOYSA-N

Application

AA-CW236 has been used as an inhibitor of m6G DNA methyltransferase in mouse embryo fibroblasts.[1]

Biochem/physiol Actions

AA-CW236 is a cell-permeable chloromethyl triazole (CMT) derivative that acts as a non-pseudosubstrate inhibitor against human O(6)-alkylguanine DNA methyltransferase (MGMT) by targeting MGMT active site Cys145 for covalent modification (KI = 24 nM, kinact = 0.03/min), displaying little affinity toward cysteines from other cellular proteins. AA-CW236 pretreatment (1 μM) effectively prevents MGMT from repairing DNA damage, causing significantly more upregulated O6-alkylguanine accumulation than the pseudosubstrate inhibitor Lomeguatrib (1 μM) when co-administered with the DNA-alkylating agent Temozolomide/TMZ (300 μM) in MCF-7 cells. Likewise, AA-CW236 is shown to boost TMZ toxicity in Caco-2 cultures (IC50 = 227 and 673 μM, respectively, with or without 3 μM AA-CW236 co-treatment).
AA-CW236 is a cell-permeable chloromethyl triazole (CMT) derivative that acts as a non-pseudosubstrate inhibitor against human O(6)-alkylguanine DNA methyltransferase (MGMT).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pennapa Thongararm et al.
Chemical research in toxicology, 33(2), 625-633 (2019-12-17)
DNA methylating agents are abundant in the environment and are sometimes used in cancer chemotherapy. They react with DNA to form methyl-DNA adducts and byproduct lesions that can be both toxic and mutagenic. Foremost among the mutagenic lesions is O6-methylguanine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service