Skip to Content
Merck
All Photos(1)

Key Documents

SML1693

Sigma-Aldrich

Pz-1

≥98% (HPLC)

Synonym(s):

N-(5-(Tert-Butyl)isoxazol-3-yl)-2-(4-(5-(1-methyl-1H-pyrazol-4-yl)-1H-benzo[d]imidazol-1-yl)phenyl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$115.00

$115.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$115.00

About This Item

Empirical Formula (Hill Notation):
C26H26N6O2
CAS Number:
Molecular Weight:
454.52
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$115.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CN1N=CC(C(C=C2)=CC3=C2N(C4=CC=C(CC(NC5=NOC(C(C)(C)C)=C5)=O)C=C4)C=N3)=C1

InChI

1S/C26H26N6O2/c1-26(2,3)23-13-24(30-34-23)29-25(33)11-17-5-8-20(9-6-17)32-16-27-21-12-18(7-10-22(21)32)19-14-28-31(4)15-19/h5-10,12-16H,11H2,1-4H3,(H,29,30,33)

InChI key

NJLMIILZNLZZFW-UHFFFAOYSA-N

Biochem/physiol Actions

Pz-1 is a cell-permeable, non-cytotoxic (up to 0.1 μM and 6 days; NIH/3T3 cells), and highly potent type II kinase inhibitor that potently inhibits RET and VEGFR2 tyrosine kinase activity (IC50 <1 nM; [ATP] = 190 μM) by targeting the kinases in their inactive DFG-out conformation, displaying significant affinity (Kd <50 nM) toward only TRKB, TRKC, GKA, FYN, SRC, TAK1, MUSK from a panel of 96 other kinases. Pz-1 inhibits cellular RET mutants (C634R, C634Y, V804L, V804M, M918T) and VEGFR2 phosphorylations, and selectively inhibits the growth of RET C634Y- than HRas G12V-transformed NIH/3T3 cultures (IC50 = 0.5 vs. 34.4 nM). Likewise, Pz-1 completely abrogated tumor formation from RET C634Y-transformed NIH/3T3 cells in mice in vivo (1 mg/kg/d, p.o.), while only up to 70% tumor suppression of HRas G12V-transformed cells was achieved with 10 mg/kg daily oral dosage.
Pz-1 is a cell-permeable, non-cytotoxic and highly potent type II kinase inhibitor that potently inhibits RET and VEGFR2 tyrosine kinase activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service