Skip to Content
Merck
All Photos(1)

Documents

SML1669

Sigma-Aldrich

NCX 4016

≥98% (HPLC)

Synonym(s):

2-(Acetyloxy)-benzoic acid 3-[(nitrooxy)methyl]phenyl ester, 2-Acetoxybenzoic acid 3-nitrooxymethylphenyl ester, 3-(Nitroxymethyl)phenyl 2-acetoxybenzoate, NCX-4016, Nitroaspirin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO7
CAS Number:
Molecular Weight:
331.28
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

−20°C

SMILES string

CC(OC1=CC=CC=C1C(OC2=CC(CO[N+]([O-])=O)=CC=C2)=O)=O

InChI

1S/C16H13NO7/c1-11(18)23-15-8-3-2-7-14(15)16(19)24-13-6-4-5-12(9-13)10-22-17(20)21/h2-9H,10H2,1H3

InChI key

IOJUJUOXKXMJNF-UHFFFAOYSA-N

Biochem/physiol Actions

NCX 4016 (nitroaspirin) is nitroderivative of aspirin that combines cyclooxygenase inhibitor with an NO donor. NCX 4016 improves postischemic ventricular dysfunction and to reduce myocardial infarct size in rabbit.
NCX-4016 can prevent the progression of cisplatin-resistant human ovarian cancer xenografts in mice. It can also regulate the expression of B-cell lymphoma 2 (Bcl-2) proteins in ovarian cancer cells. Hence, NCX-4016 is considered a promising therapeutic option against carcinoma.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karuppaiyah Selvendiran et al.
Cell cycle (Georgetown, Tex.), 7(1), 81-88 (2008-01-17)
We have previously reported the inhibitory effect of NCX-4016, a nitro derivative of aspirin, on the proliferation of cisplatin-resistant human ovarian cancer cells, in vitro (Bratasz et al., Proc Natl Acad Sci USA 2006; 103:3914-9). In this report we present

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service