Skip to Content
Merck
All Photos(1)

Key Documents

SML1667

Sigma-Aldrich

Arformoterol tartrate

≥98% (HPLC)

Synonym(s):

N-[2-Hydroxy-5-[(1R)-1-hydroxy-2-[[(1R)-2-(4-methoxyphenyl)-1-methylethyl]amino]ethyl]phenyl]-formamide (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00
25 MG
$547.00

$135.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$135.00
25 MG
$547.00

About This Item

Empirical Formula (Hill Notation):
C19H24N2O4 · C4H6O6
CAS Number:
Molecular Weight:
494.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$135.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -26 to -32°, c = 1 in H2O

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C[C@@H](NC[C@H](O)C1=CC=C(O)C(NC=O)=C1)CC2=CC=C(OC)C=C2.O[C@@H](C(O)=O)[C@H](C(O)=O)O

InChI

1S/C19H24N2O4.C4H6O6/c1-13(9-14-3-6-16(25-2)7-4-14)20-11-19(24)15-5-8-18(23)17(10-15)21-12-22;5-1(3(7)8)2(6)4(9)10/h3-8,10,12-13,19-20,23-24H,9,11H2,1-2H3,(H,21,22);1-2,5-6H,(H,7,8)(H,9,10)/t13-,19+;1-,2-/m11/s1

InChI key

FCSXYHUNDAXDRH-OKMNHOJOSA-N

Biochem/physiol Actions

Arformoterol is the active isomer of formoterol.
Arformoterol is the active isomer of formoterol. Arformoterol is a selective β2-adrenergic receptor agonist, a long-acting beta adrenergic agonist (LABA). Arformoterol is a bronchodilator used clinically as an anti-asthmatic and for treatment of chronic obstructive pulmonary disease (COPD).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service