Skip to Content
Merck
All Photos(1)

Key Documents

SML1662

Sigma-Aldrich

Buparvaquone

≥98% (HPLC)

Synonym(s):

1,4-Naphthalenedione,2-[[4-(1,1-dimethylethyl)cyclohexyl]methyl]-3-, Butalex, UNII-0354RT7LG4, 2-[(4-tert-Butylcyclohexyl)methyl]-3-hydroxy-1,4-naphthoquinone, 2-[[4-(1,1-Dimethylethyl)cyclohexyl]methyl]-3-hydroxy-1,4-naphthalenedione

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$260.00

$260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$260.00

About This Item

Empirical Formula (Hill Notation):
C21H26O3
CAS Number:
Molecular Weight:
326.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$260.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

solubility

DMSO: soluble
chloroform: soluble

storage temp.

2-8°C

SMILES string

CC(C)(C)C1CCC(CC1)CC2=C(O)C(=O)c3ccccc3C2=O

InChI

1S/C21H26O3/c1-21(2,3)14-10-8-13(9-11-14)12-17-18(22)15-6-4-5-7-16(15)19(23)20(17)24/h4-7,13-14,24H,8-12H2,1-3H3

InChI key

KLLIVCPQDTYMLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Buparvaquone is a second-generation hydroxynaphthoquinone antiprotozoal drug related to parvaquone and atovaquone.[1][2] Buparvaquone is indicated for the therapy and prophylaxis of all forms of Theileriosis[3] and found to have an anti-Leishmanial activity.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhar, S., et al.
The Veterinary Record, 119, 635-636 (1986)
Hudson, A.T., et al.
Parasitoloty, 90, 45-55 (1985)
Garnier, T., et al.
The Journal of Antimicrobial Chemotherapy, 60, 802-810 (2007)
McHardy, N., et al.
Research in Veterinary Science, 39, 29-33 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service