Skip to Content
Merck
All Photos(1)

Documents

SML1656

Sigma-Aldrich

CBR-5884

≥98% (HPLC)

Synonym(s):

5-[(2-Furanylcarbonyl)amino]-3-methyl-4-thiocyanato-2-thiophenecarboxylic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12N2O4S2
CAS Number:
Molecular Weight:
336.39
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

paste

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(C(OCC)=O)SC(NC(C2=CC=CO2)=O)=C1SC#N

InChI

1S/C14H12N2O4S2/c1-3-19-14(18)11-8(2)10(21-7-15)13(22-11)16-12(17)9-5-4-6-20-9/h4-6H,3H2,1-2H3,(H,16,17)

InChI key

QBVIRPJBDIZKBC-UHFFFAOYSA-N

Application

CBR-5884 has been used as a 3-phosphoglycerate dehydrogenase (PHGDH) inhibitor to explore the role of PHGDH in Müller cells from different retinal regions and also used in trypan blue exclusion assay to determine the number of viable MYCN cells.

Biochem/physiol Actions

CBR-5884 is a cell penetrant, potent and selective noncompetitive inhibitor of 3-phosphoglycerate dehydrogenase (PHGDH) that inhibits proliferation of melanoma and breast cancer lines. CBR-5884 inhibits de novo serine synthesis in in cancer cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optimal LC-MS metabolomic profiling reveals emergent changes to monocyte metabolism in response to lipopolysaccharide.
Leacy, et al.
Frontiers in Immunology, 14, 1116760-1116760 (2023)
Lucas D Sanor et al.
eLife, 9 (2020-01-29)
Axolotls and other salamanders can regenerate entire limbs after amputation as adults, and much recent effort has sought to identify the molecular programs controlling this process. While targeted mutagenesis approaches like CRISPR/Cas9 now permit gene-level investigation of these mechanisms, genetic
Ting Zhang et al.
eLife, 8 (2019-05-01)
The human macula is more susceptible than the peripheral retina to developing blinding conditions such as age-related macular degeneration, diabetic retinopathy. A key difference between them may be the nature of their Müller cells. We found primary cultured Müller cells
Yingfeng Xia et al.
Cancer research, 79(15), 3837-3850 (2019-05-16)
MYCN amplification drives the development of neuronal cancers in children and adults. Given the challenge in therapeutically targeting MYCN directly, we searched for MYCN-activated metabolic pathways as potential drug targets. Here we report that neuroblastoma cells with MYCN amplification show
Samah Elsaadi et al.
Experimental hematology & oncology, 10(1), 3-3 (2021-01-06)
Multiple myeloma (MM) is a hematological malignancy characterized by the clonal expansion of plasma cells in the bone marrow. To date, this disease is still incurable and novel therapeutic approaches are required. Phosphoglycerate dehydrogenase (PHGDH) is the first and rate-limiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service