Skip to Content
Merck
All Photos(1)

Key Documents

SML1646

Sigma-Aldrich

ZQ-16

≥98% (HPLC)

Synonym(s):

2-(Hexylthio)-6-hydroxy-4(3H)-pyrimidinone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$135.00

$135.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$135.00

About This Item

Empirical Formula (Hill Notation):
C10H16N2O2S
CAS Number:
Molecular Weight:
228.31
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$135.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CCCCCCSC1=NC(O)=CC(O)=N1

InChI

1S/C10H16N2O2S/c1-2-3-4-5-6-15-10-11-8(13)7-9(14)12-10/h7H,2-6H2,1H3,(H2,11,12,13,14)

InChI key

UYXOAKRMLQKLQX-UHFFFAOYSA-N

Biochem/physiol Actions

ZQ-16 is a potent and selective agonist for medium-chain free fatty acid receptor GPR84 (G protein-coupled receptor 84). ZQ-16 activates GPR84-mediated calcium mobilization, inhibition of cAMP accumulation, phosphorylation of ERK1/2, receptor desensitization and internalization, and receptor-β-arrestin interaction.
ZQ-16 is a potent and selective agonist for medium-chain free fatty acid receptor GPR84.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service