Skip to Content
Merck
All Photos(1)

Key Documents

SML1611

Sigma-Aldrich

Lafutidine

≥98% (HPLC)

Synonym(s):

(Z)-2-((Furan-2-ylmethyl)sulfinyl)-N-(4-((3-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, 2-[(2-Furanylmethyl)sulfinyl]-N-[(2Z)-4-[[4-(1-piperidinylmethyl)-2-pyridinyl]oxy]-2-buten-1-yl]-acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$118.00
50 MG
$233.00

$118.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$118.00
50 MG
$233.00

About This Item

Empirical Formula (Hill Notation):
C22H29N3O4S
CAS Number:
Molecular Weight:
431.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$118.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(CS(CC1=CC=CO1)=O)NC/C=C\COC2=NC=CC(CN3CCCCC3)=C2

InChI

1S/C22H29N3O4S/c26-21(18-30(27)17-20-7-6-14-28-20)23-9-2-5-13-29-22-15-19(8-10-24-22)16-25-11-3-1-4-12-25/h2,5-8,10,14-15H,1,3-4,9,11-13,16-18H2,(H,23,26)/b5-2-

InChI key

KMZQAVXSMUKBPD-DJWKRKHSSA-N

Biochem/physiol Actions

Lafutidine is a second generation H2 histamine receptor antagonist.
Lafutidine is a second generation H2 histamine receptor antagonist. Lafutidine has been used clinically to treat ulcers and gastro-esophageal reflux disease (GERD). Lafutidine has also been investigated as a treatment against peripheral neuropathy and for its protective stomach effects during cancer chemotherapies and long term ibuprofen use.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service