Skip to Content
Merck
All Photos(1)

Key Documents

SML1570

Sigma-Aldrich

Arglabin

≥98% (HPLC)

Synonym(s):

(3aR,4aS,6aS,9aS,9bR)-5,6,6a,7,9a,9b-Hexahydro-1,4a-dimethyl-7-methylene-3H-oxireno[8,8a]azuleno[4,5-b]furan-8(4aH)-one, 1(R),10(S)-Epoxy-5(S),5(S),7(S)-guaia-3(4),11(13)-dien-6,12-olide

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$115.00
5 MG
$465.00

$115.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
1 MG
$115.00
5 MG
$465.00

About This Item

Empirical Formula (Hill Notation):
C15H18O3
CAS Number:
Molecular Weight:
246.30
UNSPSC Code:
12352200
NACRES:
NA.77

$115.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


biological source

semisynthetic (from parthenolide)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

storage temp.

−20°C

SMILES string

O1[C@@]32[C@@]1(CC[C@@H]4[C@H](OC(=O)C4=C)[C@@H]3C(=CC2)C)C

InChI

1S/C15H18O3/c1-8-4-7-15-11(8)12-10(9(2)13(16)17-12)5-6-14(15,3)18-15/h4,10-12H,2,5-7H2,1,3H3/t10-,11-,12-,14-,15+/m0/s1

InChI key

UVJYAKBJSGRTHA-ZCRGAIPPSA-N

Biochem/physiol Actions

Arglabin is a natural product with antitumor and anti-inflammatory activity. Arglabin competitively inhibits the binding of farnesyl diphosphate to farnesyl transferase (FTase), preventing the activation of RAS proto-oncogene by preventing the incorporation of farnesyl pyrophosphate into human H-ras proteins. Arglabin is also an inhibitor of the NLRP3 inflammasome. Arglabin reduces inflammation and plasma lipids and has shown a marked reduction in atherosclerotic lesions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Liu et al.
OncoTargets and therapy, 13, 2047-2055 (2020-03-27)
Epithelial membrane protein 1 (EMP1), a member of the EMP family, is overexpressed in a large number of tumors and is thought to be a cellular connexin on the cell membrane and is involved in proliferation, invasion, metastasis of tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service