Skip to Content
Merck
All Photos(1)

Key Documents

SML1530

Sigma-Aldrich

SB-334867

≥98% (HPLC)

Synonym(s):

N-(2-Methyl-6-benzoxazolyl)-N′-1,5-naphthyridin-4-yl-urea, SB-334867, SB-334867A, SB334867

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$168.00
25 MG
$682.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$168.00
25 MG
$682.00

About This Item

Empirical Formula (Hill Notation):
C17H13N5O2
CAS Number:
Molecular Weight:
319.32
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$168.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

O=C(NC1=CC=C(N=C(C)O2)C2=C1)NC3=CC=NC4=C3N=CC=C4

InChI

1S/C17H13N5O2/c1-10-20-12-5-4-11(9-15(12)24-10)21-17(23)22-14-6-8-18-13-3-2-7-19-16(13)14/h2-9H,1H3,(H2,18,21,22,23)

InChI key

AKMNUCBQGHFICM-UHFFFAOYSA-N

Biochem/physiol Actions

Lateral hypothalamic orexin neuron activation leads to the development of morphine conditioned place preference, which is preferably blocked by SB-334867.[1]
SB-334867 is a non-peptide selective antagonist of the orexin OX1 receptor.
SB-334867 is a non-peptide selective antagonist of the orexin OX1 receptor. SB-334867 blocks orexin-A induced calcium response with a pKb of 7.23. SB-334867 blocks increases in heart rate and body temperature induced by administratin of methamphetamine, and can inhibit the development of tolerance to the analgesic properties of morphine in rodent models. SB-334867 also shows anxiolytic and sedative effects.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential effects of the hypocretin 1 receptor antagonist SB 334867 on high?fat food self?administration and reinstatement of food seeking in rats.
Nair S G, et al.
British Journal of Pharmacology, 154(2), 406-416 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service