Skip to Content
Merck
All Photos(1)

Key Documents

SML1485

Sigma-Aldrich

SW033291

≥98% (HPLC)

Synonym(s):

2-(Butane-1-sulfinyl)-4-phenyl-6-thiophen-2-yl-thieno[2,3-b]pyridin-3-ylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$229.00
25 MG
$731.00

$229.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$229.00
25 MG
$731.00

About This Item

Empirical Formula (Hill Notation):
C21H20N2OS3
CAS Number:
Molecular Weight:
412.59
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$229.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to yellow-green

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

NC1=C(S(CCCC)=O)SC2=NC(C3=CC=CS3)=CC(C4=CC=CC=C4)=C21

InChI

1S/C21H20N2OS3/c1-2-3-12-27(24)21-19(22)18-15(14-8-5-4-6-9-14)13-16(23-20(18)26-21)17-10-7-11-25-17/h4-11,13H,2-3,12,22H2,1H3

InChI key

LCYAYKSMOVLVRL-UHFFFAOYSA-N

Biochem/physiol Actions

SW033291 is an inhibitor of the prostaglandin-degrading enzyme 15-PGDH
SW033291 is an inhibitor of the prostaglandin-degrading enzyme 15-PGDH. PGE2 is known to promote hematopoiesis and to promote reeneration by causing stem cells to proliferate. 15-PGDH degrades prostaglandin PGE2 and is a negative regulator of tissue regeneration and repair. SW033291 inhibition of 15-PGDH doubled levels of PGE2 and doubled rates of colon and liver tissue regeneration. Mice given SW033291 after receiving a bone marrow transplant had a two- to threefold increase in the number of donor cells that homed to the their bone marrow.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yichao Zhang et al.
iScience, 24(9), 102983-102983 (2021-09-07)
Rosiglitazone, a synthetic peroxisome proliferator-activated receptor γ (PPARγ) ligand, has been reported to reduce growth hormone (GH) and insulin-like growth factor-1 (IGF-1) in 10 patients with acromegaly. However, the mechanisms remain unknown. Here, we reveal that PPARγ directly enhances 15-hydroxyprostaglandin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service