Skip to Content
Merck
All Photos(1)

Key Documents

SML1475

Sigma-Aldrich

Ampiroxicam

≥98% (HPLC)

Synonym(s):

CP 65703, Carbonic acid ethyl 1 -[[2-methyl-1,1-dioxido-3-[(2-pyridinylamino)carbonyl]-2H-1,2-benzothiazin-4-yl]oxy]ethyl ester, Carbonic acid ethyl 1-[[2-methyl-3-[(2-pyridinylamino)carbonyl]-2H-1,2-benzothiazin-4-yl]oxy]ethyl ester S,S-dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$97.50

$97.50


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$97.50

About This Item

Empirical Formula (Hill Notation):
C20H21N3O7S
CAS Number:
Molecular Weight:
447.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$97.50


Estimated to ship onMay 25, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

CCOC(OC(OC1=C(C(NC2=CC=CC=N2)=O)N(C)S(=O)(C3=CC=CC=C31)=O)C)=O

InChI

1S/C20H21N3O7S/c1-4-28-20(25)30-13(2)29-18-14-9-5-6-10-15(14)31(26,27)23(3)17(18)19(24)22-16-11-7-8-12-21-16/h5-13H,4H2,1-3H3,(H,21,22,24)

InChI key

LSNWBKACGXCGAJ-UHFFFAOYSA-N

Biochem/physiol Actions

Ampiroxicam is a non-steroidal anti-inflammatory drug (NSAID), a prodrug of Cox 1/2 inhibitor piroxicam.
Ampiroxicam is an ether carbonate prodrug.This nonacidic drug does not have the ability to prevent detectable prostaglandin production in vitro.[1]
Antiinflammatory NSAID, prodrug of piroxicam

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Carty et al.
Agents and actions, 39(3-4), 157-165 (1993-07-01)
Ampiroxicam is a nonacidic ether carbonate prodrug of piroxicam. Our results demonstrate that, in contrast to piroxicam, ampiroxicam does not possess detectable prostaglandin synthesis inhibitory activity in vitro. Ampiroxicam, however, has similar in vivo potency to piroxicam in suppressing paw

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service