Skip to Content
Merck
All Photos(1)

Key Documents

SML1460

Sigma-Aldrich

SAMS Peptide trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

HMRSAMSGLHLVKRR trifluoroacetate salt, His-Met-Arg-Ser-Ala-Met-Ser-Gly-Leu-His-Leu-Val-Lys-Arg-Arg-OH trifluoroacetate salt, L-Histidyl-L-methionyl-L-arginyl-L-seryl-L-alanyl-L-methionyl-L-seryl-glycyl-L-leucyl-L-histidyl-L-leucyl-L-valyl-L-lysyl-L-arginyl-L-arginine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$204.00

$204.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C74H131N29O18S2 · xC2HF3O2
CAS Number:
Molecular Weight:
1779.15 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$204.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)Cc2nc[nH]c2)CCSC)CCCNC(=N)N)CO)C)C(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc1nc[nH]c1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCCNC(

InChI

1S/C74H131N29O18S2/c1-38(2)27-51(66(115)100-53(30-43-32-84-37-90-43)67(116)99-52(28-39(3)4)68(117)103-57(40(5)6)70(119)97-45(15-10-11-21-75)61(110)95-46(16-12-22-85-72(77)78)62(111)98-50(71(120)121)18-14-24-87-74(81)82)92-56(106)33-88-60(109)54(34-104)101-65(114)48(19-25-122-8)93-58(107)41(7)91-69(118)55(35-105)102-63(112)47(17-13-23-86-73(79)80)96-64(113)49(20-26-123-9)94-59(108)44(76)29-42-31-83-36-89-42/h31-32,36-41,44-55,57,104-105H,10-30,33-35,75-76H2,1-9H3,(H,83,89)(H,84,90)(H,88,109)(H,91,118)(H,92,106)(H,93,107)(H,94,108)(H,95,110)(H,96,113)(H,97,119)(H,98,111)(H,99,116)(H,100,115)(H,101,114)(H,102,112)(H,103,117)(H,120,121)(H4,77,78,85)(H4,79,80,86)(H4,81,82,87)/t41-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,57-/m0/s1

InChI key

YJAHUEAICBOBTC-CSVPVIMBSA-N

General description

SAMS Peptide trifluoroacetate salt is a peptide having a putative AMPK phosphorylation site (Ser568) of carbohydrate-responsive element-binding protein (ChREBP) and a peptide containing a cAMP-dependent protein kinase (PKA) phosphorylation site (Thr666).[1]

Biochem/physiol Actions

SAMS Peptide is a synthetic substrate selective for AMP-activated protein kinase (AMPK). Its sequence is based on acetyl-CoA carboxylase, which is phosphorylated exclusively by the AMPK, but with serine (Ser)-79 replaced by alanine SAMS peptide is more specific for AMPK than acetyl-CoA carboxylase itself.
Substrate selecive for AMP-activated protein kinase (AMPK)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism for fatty acid ?sparing? effect on glucose-induced transcription regulation of carbohydrate-responsive element-binding protein by AMP-activated protein kinase
Kawaguchi, et al.
Test, 277(6), 3829-3835 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service