Skip to Content
Merck
All Photos(1)

Key Documents

SML1455

Sigma-Aldrich

DAA-I acetate salt

≥98% (HPLC)

Synonym(s):

5-L-Isoleucine-2-10-Angiotensin I acetate salt, Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu acetate, RVYIHPFHL acetate, [Des-Asp1-Ile5]angiotensin I, des-Asp-angiotensin I acetate, des-aspartate-angiotensin-I acetate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$209.00

$209.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$209.00

About This Item

Empirical Formula (Hill Notation):
C58H84N16O11 · xC2H4O2
CAS Number:
Molecular Weight:
1181.39 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$209.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

film

color

colorless

shipped in

wet ice

storage temp.

−20°C

Biochem/physiol Actions

DAA-I (des-aspartate-angiotensin-I) is an agonist on the angiotensin AT1 receptor and releases prostaglandins which mediate its actions. DAA-I administer at doses lesser than Km of metabolizing enzymes antagonize the deleterious actions of angiotensin II. DAA-I appear function in vivo as a physiological antagonist to angiotensin II.
DAA-I (des-aspartate-angiotensin-I) is an agonist on the angiotensin AT1 receptor.
Des-aspartate-angiotensin I (DAA-I) is a nine-amino acid angiotensin peptide[1] and a metabolite of angiotensin[2] DAA-I mediates attenuation of early inflammatory processes[1] and intercellular adhesion molecule-1 (ICAM-1) formation in animal models.[3]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abd Jalil Rufaihah et al.
Life sciences, 78(12), 1341-1351 (2006-01-21)
We investigate the influence of des-Aspartate-angiotensin-I (DAA-I) on the cytokine expression profile in a rodent model of myocardial infarction. Myocardial infarction model was created in female Wistar rats by coronary artery ligation. Animals were randomized to receive intravenously either a
Qiang Wen et al.
European journal of pharmacology, 658(2-3), 193-199 (2011-03-05)
The present study investigated the actions of des-aspartate-angiotensin I (DAA-I) on infarct size and three early inflammatory events in acute myocardial ischemia-reperfusion injury in rats. The rationale was based on earlier findings showing that chronic daily administration of DAA-I attenuated
Yong-Chiat Wong et al.
Biochemical pharmacology, 82(9), 1198-1208 (2011-08-02)
Although clinical studies suggested that blockade of the renin-angiotensin system may prevent diabetes, the mechanism is uncertain. As a follow-up to an earlier study, we investigated how des-aspartate-angiotensin-1 (DAA-1) and its metabolite, angiotensin IV (Ang-IV) improved glucose tolerance in diet-induced
Meng-Kwoon Sim
European journal of pharmacology, 760, 36-41 (2015-04-22)
The review describes DAA-I (des-aspartate-angiotensin-I) as a prototype of a novel class of drugs that acts as agonists on the angiotensin AT1 receptor or ARAs (angiotensin receptor agonists). DAA-I is a component of the renin angiotensin system. Earlier studies showed
M Dharmani et al.
Regulatory peptides, 129(1-3), 213-219 (2005-06-02)
The present study investigated the action of des-aspartate-angiotensin I (DAA-I) on the pressor action of angiotensin II in the renal and mesenteric vasculature of WKY, SHR and streptozotocin (STZ)-induced diabetic rats. Angiotensin II-induced a dose-dependent pressor response in the renal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service