Skip to Content
Merck
All Photos(1)

Key Documents

SML1446

Sigma-Aldrich

HTH-01-015

≥98% (HPLC)

Synonym(s):

5,13-Dihydro-4,5,13-trimethyl-2-[[1-(4-piperidinyl)-1H-pyrazol-4-yl]amino]-6H-naphtho[2,3-e]pyrimido[5,4-b][1,4]diazepin-6-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$139.00
25 MG
$555.00

$139.00


Estimated to ship onMay 28, 2025



Select a Size

Change View
5 MG
$139.00
25 MG
$555.00

About This Item

Empirical Formula (Hill Notation):
C26H28N8O
CAS Number:
Molecular Weight:
468.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$139.00


Estimated to ship onMay 28, 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CC1=C2C(N(C)C(C=C(C=CC=C3)C3=C4)=C4C(N2C)=O)=NC(NC5=CN(C6CCNCC6)N=C5)=N1

InChI

1S/C26H28N8O/c1-16-23-24(31-26(29-16)30-19-14-28-34(15-19)20-8-10-27-11-9-20)32(2)22-13-18-7-5-4-6-17(18)12-21(22)25(35)33(23)3/h4-7,12-15,20,27H,8-11H2,1-3H3,(H,29,30,31)

InChI key

CHSDJDLAKKAWCI-UHFFFAOYSA-N

Biochem/physiol Actions

HTH-01-015 is a potent and selective inhibitor of NUAK1 kinase, a member of the AMPK family that are that are activated by the LKB1 (liver kinaseB1) tumour suppressor kinase and involved in development and proliferation.
HTH-01-015 is a potent and selective inhibitor of NUAK1 kinase, a member of the AMPK family that are that are activated by the LKB1 (liver kinaseB1) tumour suppressor kinase and involved in development and proliferation. HTH-01-015 has an IC50 of 100 nM for NUAK1, >100-fold higher than NUAK2, and has no significant inhibition of 139 other kinases tested, including ten AMPK-related kinase family members. Treatment with HTH-01-015 was found to suppress proliferation, restricting cells from entering into mitosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service