Skip to Content
Merck
All Photos(1)

Key Documents

SML1391

Sigma-Aldrich

Olmesartan medoxomil

≥98% (HPLC)

Synonym(s):

4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic acid (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl ester, CS-866

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$144.00
250 MG
$564.00

$144.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$144.00
250 MG
$564.00

About This Item

Empirical Formula (Hill Notation):
C29H30N6O6
CAS Number:
Molecular Weight:
558.59
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$144.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CCCC1=NC(C(O)(C)C)=C(C(OCC2=C(C)OC(O2)=O)=O)N1CC(C=C3)=CC=C3C4=CC=CC=C4C5=NN=NN5

InChI

1S/C29H30N6O6/c1-5-8-23-30-25(29(3,4)38)24(27(36)39-16-22-17(2)40-28(37)41-22)35(23)15-18-11-13-19(14-12-18)20-9-6-7-10-21(20)26-31-33-34-32-26/h6-7,9-14,38H,5,8,15-16H2,1-4H3,(H,31,32,33,34)

InChI key

UQGKUQLKSCSZGY-UHFFFAOYSA-N

Gene Information

human ... AGTR1(185)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Olmesartan medoxomil is a selective Angiotensin II Type I receptor blocker and antihypertensive drug. Olmesartan medoxomil is converted enzymatically to the active form olmesartan.
Olmesartan medoxomil is considered as an antihypertensive agent. It is an angiotensin 2 type 1 antagonist receptor.[1] It is a prodrug which releases the parent compound olmesartan after ingestion. Olmesartan is ejected via kidney and liver.[2] In humans, olmesartan protects renal and cardiovascular systems. This drug may be used in the treatment of hypertension.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Angiotensin II Inhibitors: Advances in Research and Application (2012)
Hans R Brunner
Journal of human hypertension, 16 Suppl 2, S13-S16 (2002-04-23)
The new orally active angiotensin II (A II) type-1 receptor antagonist olmesartan medoxomil is a prodrug, which is rapidly converted in vivo to the active metabolite, olmesartan. The pharmacology, antihypertensive efficacy and safety of olmesartan medoxomil and/or the pharmacologically active
Case Studies in Modern Drug Discovery and Development (2012)
Current Cardiovascular Drugs (2005)
Linear mixed-effects model of QTc prolongation for olmesartan medoxomil.
SaeHeum Song et al.
Journal of clinical pharmacology, 56(1), 96-100 (2015-06-23)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service