Skip to Content
Merck
All Photos(1)

Key Documents

SML1390

Sigma-Aldrich

Fexaramine

≥98% (HPLC)

Synonym(s):

3-[3-[(Cyclohexylcarbonyl)[[4′-(dimethylamino)[1,1′-biphenyl]-4-yl]methyl]amino]phenyl]-2-propenoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$184.00
25 MG
$726.00

$184.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$184.00
25 MG
$726.00

About This Item

Empirical Formula (Hill Notation):
C32H36N2O3
CAS Number:
Molecular Weight:
496.64
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

$184.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN(C)C(C=C1)=CC=C1C2=CC=C(CN(C3=CC=CC(/C=C/C(OC)=O)=C3)C(C4CCCCC4)=O)C=C2

InChI

1S/C32H36N2O3/c1-33(2)29-19-17-27(18-20-29)26-15-12-25(13-16-26)23-34(32(36)28-9-5-4-6-10-28)30-11-7-8-24(22-30)14-21-31(35)37-3/h7-8,11-22,28H,4-6,9-10,23H2,1-3H3/b21-14+

InChI key

VLQTUNDJHLEFEQ-KGENOOAVSA-N

General description

Fexaramine belongs to the nuclear hormone receptor family.[1]

Application

Fexaramine has been used to study its action on RANKL (receptor activator of nuclear factor-κB ligand)-induced osteoclastogenesis in mouse model.[1]

Biochem/physiol Actions

Fexaramine is a potent and selective agonist of the bile acid sensor farnesoid X receptor (FXR) in the gut with an EC50 of 25 nM and no activity found for other nuclear receptors. Fexaramine induces enteric fibroblast growth factor 15 (FGF15), causing alterations in bile acid composition without activating FXR target genes in the liver. In mouse studies, fexaramine enhanced thermogenesis and browning of white adipose tissue while reducing diet-induced weight gain, body-wide inflammation and hepatic glucose production. Fexaramine also improved insulin responsiveness.
Fexaramine is an agonist of the farnesoid X receptor (FXR).
Fexaramine might regulate lipid and glucose metabolism and can serve as a therapeutic target in the treatment of fatty liver disease, type 2 diabetes and obesity.[2] Fexaramine might mediate cholesterol homeostasis and promotes osteoblast differentiation and suppresses differentiation of osteoclast.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Farnesoid X receptor modulators (2011?2014): a patent review.
Sepe V, et al.
Expert Opinion on Therapeutic Patents, 25(8), 885-896 (2015)
Fexaramine Inhibits Receptor Activator of Nuclear Factor-?B Ligand-induced Osteoclast Formation via Nuclear Factor of Activated T Cells Signaling Pathways.
Zheng T, et al.
Journal of bone metabolism, 24(4), 207-215 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service