Skip to Content
Merck
All Photos(1)

Key Documents

SML1387

Sigma-Aldrich

Mps1-IN-2

≥98% (HPLC)

Synonym(s):

9-Cyclopentyl-2-[[2-ethoxy-4-(4-hydroxy-1-piperidinyl)phenyl]amino]-5,7,8,9-tetrahydro-5-methyl-6H-pyrimido[4,5-b][1,4]diazepin-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H36N6O3
CAS Number:
Molecular Weight:
480.60
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 1 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

OC(CC1)CCN1C2=CC=C(NC3=NC(N(C4CCCC4)CCC(N5C)=O)=C5C=N3)C(OCC)=C2

InChI

1S/C26H36N6O3/c1-3-35-23-16-19(31-13-10-20(33)11-14-31)8-9-21(23)28-26-27-17-22-25(29-26)32(18-6-4-5-7-18)15-12-24(34)30(22)2/h8-9,16-18,20,33H,3-7,10-15H2,1-2H3,(H,27,28,29)

InChI key

WELBJLUKWAJOQV-UHFFFAOYSA-N

Biochem/physiol Actions

Mps1-IN-2 is a potent and selective inhibitor of Mps1 kinase.
Mps1-IN-2 is a potent and selective inhibitor of Mps1 kinase. Mps1-IN-2 also significantly inhibits Plk1.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service