Skip to Content
Merck
All Photos(1)

Key Documents

SML1385

Sigma-Aldrich

Lomitapide

≥98% (HPLC)

Synonym(s):

AEGR-733, BMS-201038, N-(2,2,2-Trifluoroethyl)-9-[4-[4-[[[4′-(trifluoromethyl)[1,1′-biphenyl]2-yl]carbonyl]amino]-1-piperidinyl]butyl]9H-fluoren-9-carboxamde

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$218.00
25 MG
$870.00

$218.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$218.00
25 MG
$870.00

About This Item

Empirical Formula (Hill Notation):
C39H37F6N3O2
CAS Number:
Molecular Weight:
693.72
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$218.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NCC(F)(F)F)C1(C(C=CC=C2)=C2C3=C1C=CC=C3)CCCCN(CC4)CCC4NC(C5=C(C6=CC=C(C(F)(F)F)C=C6)C=CC=C5)=O

InChI

1S/C39H37F6N3O2/c40-38(41,42)25-46-36(50)37(33-13-5-3-10-30(33)31-11-4-6-14-34(31)37)21-7-8-22-48-23-19-28(20-24-48)47-35(49)32-12-2-1-9-29(32)26-15-17-27(18-16-26)39(43,44)45/h1-6,9-18,28H,7-8,19-25H2,(H,46,50)(H,47,49)

InChI key

MBBCVAKAJPKAKM-UHFFFAOYSA-N

Gene Information

Application

Lomitapide has been used as a microsomal triglyceride transfer protein (MTP) inhibitor to study its effects on very-low-density lipoproteins (VLDL) export in mouse hepatocytes.[1]

Biochem/physiol Actions

Lomitapide can lower the secretion of very-low-density lipoproteins (VLDL) and chylomicrons.[2]
Lomitapide is an inhibitor of microsomal triglyceride transfer protein (MTP).
Lomitapide is an inhibitor of microsomal triglyceride transfer protein (MTP). Lomitapide has been shown to be highly effective in reducing LDL-cholesterol and triglycerides, and has been aproved for treatment of homozygous familial hypercholesterolemia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Targeting Hepatic Glutaminase 1 Ameliorates Non-alcoholic Steatohepatitis by Restoring Very-Low-Density Lipoprotein Triglyceride Assembly
Simon J, et al.
Cell Metabolism, 31(3), 605-622 (2020)
Severe hypertriglyceridemia with pancreatitis: thirteen years? treatment with lomitapide
Sacks F M, et al.
JAMA Internal Medicine, 174(3), 443-447 (2014)
Frank M Sacks et al.
JAMA internal medicine, 174(3), 443-447 (2013-12-25)
Recurrent pancreatitis is a potentially fatal complication of severe hypertriglyceridemia. Genetic defects and lifestyle risk factors may render this condition unresponsive to current treatments. We report this first case of long-term management of intractable near-fatal recurrent pancreatitis secondary to severe
Jorge Simon et al.
Cell metabolism, 31(3), 605-622 (2020-02-23)
Non-alcoholic steatohepatitis (NASH) is characterized by the accumulation of hepatic fat in an inflammatory/fibrotic background. Herein, we show that the hepatic high-activity glutaminase 1 isoform (GLS1) is overexpressed in NASH. Importantly, GLS1 inhibition reduces lipid content in choline and/or methionine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service