Skip to Content
Merck
All Photos(1)

Key Documents

SML1357

Sigma-Aldrich

VU0453379 hydrochloride

≥98% (HPLC)

Synonym(s):

(S)2-CyclopentylN((1-isopropylpyrrolidin2-yl)-9-methyl-1-oxo-2,9-dihydro1Hpyrrido[3,4b]indole-4-carboxamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$464.00

$464.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$464.00

About This Item

Empirical Formula (Hill Notation):
C26H34N4O2 · HCl
CAS Number:
Molecular Weight:
471.03
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$464.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CN1C2=C(C(C(NC[C@@H]3CCCN3C(C)C)=O)=CN(C4CCCC4)C2=O)C5=CC=CC=C51.Cl

InChI

1S/C26H34N4O2.ClH/c1-17(2)29-14-8-11-19(29)15-27-25(31)21-16-30(18-9-4-5-10-18)26(32)24-23(21)20-12-6-7-13-22(20)28(24)3;/h6-7,12-13,16-19H,4-5,8-11,14-15H2,1-3H3,(H,27,31);1H/t19-;/m0./s1

InChI key

OCMGNMGYQPPMFF-FYZYNONXSA-N

Biochem/physiol Actions

VU0453379 is a highly selective CNS penetrant positive allosteric modulator of Glucagon-like peptide-1 (GLP-1) with GLP-1 EC50 = 1.3 μ,M, 59.2% GLP-1 max. VU0453379 potentiated low-dose exenatide to augment insulin secretion in primary mouse pancreatic islets. In a model of Parkinson′s disease it showed a significant reversal of haloperidol-induced catalepsy at 30 mg/kg.
VU0453379 is a highly selective CNS penetrant positive allosteric modulator of Glucagon-like peptide-1 (GLP-1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service