Skip to Content
Merck
All Photos(1)

Documents

SML1300

Sigma-Aldrich

MitoB

≥98% (HPLC)

Synonym(s):

3-(Dihydroxyboronyl)benzyltriphenylphosphonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23BBrO2P
CAS Number:
Molecular Weight:
477.14
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

MitoB is a mitochondria-targeted mass spectrometry probe used to measure hydrogen peroxide (H2O2) levels within mitochondria. MitoB reacts selelectively with H2O2 to form a new product, MitoP. The ratio of MitoP to MitoB can be used to determine mitochondrial H2O2 concentration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Lujing Jiang et al.
Theranostics, 11(4), 1703-1720 (2021-01-08)
Rationale: Reactive oxygen species (ROS) burst from mitochondrial complex I is considered the critical cause of ischemia/reperfusion (I/R) injury. Ginsenoside Rb1 has been reported to protect the heart against I/R injury; however, the underlying mechanism remains unclear. This work aimed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service