Skip to Content
Merck
All Photos(1)

Key Documents

SML1290

Sigma-Aldrich

PA-824

≥98% (HPLC)

Synonym(s):

(6S)- 6,7-Dihydro-2-nitro-6-[[4-(trifluoromethoxy)phenyl]methoxy]-5H-Imidazo[2,1-b][1,3]oxazine, (6S)-2-Nitro-6-{[4-(trifluoromethoxy)benzyl]oxy}-6,7-dihydro-5H-imidazo[2,1-b][1,3]oxazine, (S)-PA 824, PA 824, Pretomanid

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$306.00
50 MG
$1,230.00

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$306.00
50 MG
$1,230.00

About This Item

Empirical Formula (Hill Notation):
C14H12F3N3O5
CAS Number:
Molecular Weight:
359.26
UNSPSC Code:
12352200
NACRES:
NA.77

$306.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -40 to -48°, c = 1 in methanol

color

white to brown

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

FC(F)(F)Oc1ccc(cc1)CO[C@H]2C[n]3c(nc(c3)[N+](=O)[O-])OC2

InChI

1S/C14H12F3N3O5/c15-14(16,17)25-10-3-1-9(2-4-10)7-23-11-5-19-6-12(20(21)22)18-13(19)24-8-11/h1-4,6,11H,5,7-8H2/t11-/m0/s1

InChI key

ZLHZLMOSPGACSZ-NSHDSACASA-N

Biochem/physiol Actions

Anti-tuberculosis drug
PA-824 is a pro-drug that has an effect on both replicating and non-replicating Mycobacterium tuberculosis. It is activated by deazaflavin (cofactor F420)-dependent nitroreducase.[1]
PA-824 is an anti-tuberculosis drug that exhibits multiple mechanism of action, including both cell wall inhibition and respiratory poisoning.
PA-824 is an anti-tuberculosis drug.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The mechanism of action of PA-824: novel insights from transcriptional profiling.
Manjunatha U, et al.
Communicative & Integrative Biology, 2(3), 215-218 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service