Skip to Content
Merck
All Photos(1)

Key Documents

SML1225

Sigma-Aldrich

Indisulam

≥98% (HPLC)

Synonym(s):

N-(3-chloro-1H-indol-7-yl)-4-sulfamoylbenzenesulfonamide, N1-(3-Chloro-1H-indol-7-yl)-1,4-benzenedisulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$184.00
25 MG
$726.00

$184.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$184.00
25 MG
$726.00

About This Item

Empirical Formula (Hill Notation):
C14H12ClN3O4S2
CAS Number:
Molecular Weight:
385.85
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$184.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to pink

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

ClC1=CNC2=C1C=CC=C2NS(C3=CC=C(S(=O)(N)=O)C=C3)(=O)=O

InChI

1S/C14H12ClN3O4S2/c15-12-8-17-14-11(12)2-1-3-13(14)18-24(21,22)10-6-4-9(5-7-10)23(16,19)20/h1-8,17-18H,(H2,16,19,20)

InChI key

SETFNECMODOHTO-UHFFFAOYSA-N

Biochem/physiol Actions

Indisulam is a carbonic anhydrase inibitor and Antitumor CDK inhibitor.
Indisulam is a carbonic anhydrase inibitor and Antitumor CDK inhibitor. Indisulam targets the G1 phase of the cell cycle by depleting cyclin E. inducing p53 and p21, and inhibiting CDK2, causing a blockade in the G1/S transition.
Indisulam is synthetic sulfonamide compound.[1] In vitro and in vivo studies point out that indisulam prevents retinoblastoma protein phosphorylation. Also, it inhibits cyclins A and B1 activity. Indisulam is known to promote apoptosis. Indisulam is a potent inhibitor of cellular dehydrogenases and thus, is likely to interfere with metabolic pathways such as malate-aspartate shuttle, glycolysis and gluconeogenesis.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Semi-physiological model describing the hematological toxicity of the anti-cancer agent indisulam.
van Kesteren C, et al.
Investigational New Drugs, 23(3), 225-234 (2005)
Sung Ah Kim et al.
Molecules and cells, 43(11), 935-944 (2020-11-11)
Aryl hydrocarbon receptor nuclear translocator (ARNT) plays an essential role in maintaining cellular homeostasis in response to environmental stress. Under conditions of hypoxia or xenobiotic exposure, ARNT regulates the subset of genes involved in adaptive responses, by forming heterodimers with
Cristina Mayor-Ruiz et al.
Nature chemical biology, 16(11), 1199-1207 (2020-08-05)
Targeted protein degradation is a new therapeutic modality based on drugs that destabilize proteins by inducing their proximity to E3 ubiquitin ligases. Of particular interest are molecular glues that can degrade otherwise unligandable proteins by orchestrating direct interactions between target

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service