Skip to Content
Merck
All Photos(1)

Key Documents

SML1221

Sigma-Aldrich

Dutasteride

≥98% (HPLC)

Synonym(s):

17β-N-[2,5-Bis(trifluoromethyl)phenyl]carbamoyl-4-aza-5α-androst-1-en-3-one, (5α,17β)-N-[2,5-Bis(trifluoromethyl)phenyl]-3-oxo-4-azaandrost-1-ene-17-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$126.00
50 MG
$506.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$126.00
50 MG
$506.00

About This Item

Empirical Formula (Hill Notation):
C27H30F6N2O2
CAS Number:
Molecular Weight:
528.53
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +18 to +24°, c = 1 in chloroform-d

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C1C=C[C@@]2(C)[C@](CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@@H]4C(NC5=CC(C(F)(F)F)=CC=C5C(F)(F)F)=O)([H])N1

InChI

1S/C27H30F6N2O2/c1-24-11-9-17-15(4-8-21-25(17,2)12-10-22(36)35-21)16(24)6-7-19(24)23(37)34-20-13-14(26(28,29)30)3-5-18(20)27(31,32)33/h3,5,10,12-13,15-17,19,21H,4,6-9,11H2,1-2H3,(H,34,37)(H,35,36)/t15-,16-,17-,19+,21+,24-,25+/m0/s1

InChI key

JWJOTENAMICLJG-QWBYCMEYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Dutasteride is a potent dual inhibitor of 5α-reductase isoenzymes types 1 and 2 (IC50 = 6 nM 5-AR1; 7 nM 5-AR2). Dutasteride blocks testosterone conversion to dihydrotesterone, and is used clinically for treating benign prostatic hyperplasia (BPH).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Akira Tsujimura et al.
Urology, 85(1), 274-274 (2014-12-03)
To evaluate histologic change in human prostate samples treated with dutasteride and to elucidate direct effects of dutasteride on human prostate tissue, the present study was conducted by using a xenograft model with improved severe combined immunodeficient (super-SCID) mice, although
Christopher Wu et al.
Expert opinion on pharmacotherapy, 14(10), 1399-1408 (2013-06-12)
Benign prostatic hyperplasia (BPH) is an age-related phenomenon associated with prostatic enlargement and bladder outlet obstruction that can cause significant lower urinary tract symptoms that greatly affect quality of life. Dutasteride is a selective inhibitor of type 1 and type
5α-Reductase Inhibition Coupled with Short Off Cycles Increases Survival in the LNCaP Xenograft Prostate Tumor Model on Intermittent Androgen Deprivation Therapy.
Pascal, L.E., et al.
The Journal of Urology, Oct 31 (2014)
Sungsin Jo et al.
Arthritis research & therapy, 22(1), 121-121 (2020-05-26)
Ankylosing spondylitis (AS) is characteristically male-predominant, and progressive spinal ankylosis affects male patients more severely; however, the hormonal effects in males with AS are poorly understood. In the present study, the regulatory effects of dutasteride, a 5-α reductase inhibitor that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service